Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 47229635-47229882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZMYND8
Entrez Gene ID
23613
Gene Name
zinc finger MYND-type containing 8
Gene Aliases
PRKCBP1, PRO2893, RACK7
UniGene
Hs.446240
Gene Chromosome Location
Chr.20: 47209214-47357780 on Build GRCh38
Transcripts
NM_001281771.2 NM_001281772.2 NM_001281773.2 NM_001281774.2 NM_001281775.2 NM_001281776.2 NM_001281777.2 NM_001281778.2 NM_001281779.2 NM_001281780.2 NM_001281781.2 NM_001281782.2 NM_001281783.2 NM_001281784.2 NM_012408.5 NM_183047.3 NM_183048.3 XM_005260356.4 XM_005260358.2 XM_005260360.4 XM_005260362.2 XM_005260364.2 XM_005260366.2 XM_006723760.3 XM_006723762.3 XM_006723763.1 XM_006723766.1 XM_011528749.2 XM_011528751.1 XM_011528752.1 XM_011528753.1 XM_011528754.1 XM_017027760.1 XM_017027761.1 XM_017027762.1 XM_017027763.1 XM_017027764.1 XM_017027765.1 XM_017027766.1 XM_017027767.1 XM_017027768.1 XM_017027769.1 XM_017027770.1 XM_017027771.1 XM_017027772.1 XM_017027773.1 XM_017027774.1 XM_017027775.1 XM_017027776.1 XM_017027777.1 XM_017027778.1 XM_017027779.1 XM_017027780.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6419008 c.2738G>T p.G913V Chr.20: 47229787-47229787 on Build GRCh38
6419009 c.2897G>T p.G966V Chr.20: 47229787-47229787 on Build GRCh38
7252023 c.2951T>C p.I984T Chr.20: 47229733-47229733 on Build GRCh38
7252022 c.2792T>C p.I931T Chr.20: 47229733-47229733 on Build GRCh38
7252024 c.2870T>C p.I957T Chr.20: 47229733-47229733 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762069313 Chr.20: 47229677-47229677 on Build GRCh38
rs748922070 Chr.20: 47229841-47229841 on Build GRCh38
rs751661843 Chr.20: 47229716-47229716 on Build GRCh38
rs758926824 Chr.20: 47229690-47229690 on Build GRCh38
rs1055514930 Chr.20: 47229800-47229800 on Build GRCh38
rs200776173 Chr.20: 47229712-47229712 on Build GRCh38
rs768268744 Chr.20: 47229820-47229820 on Build GRCh38
rs3746488 Chr.20: 47229675-47229675 on Build GRCh38
rs767043217 Chr.20: 47229702-47229702 on Build GRCh38
rs756292218 Chr.20: 47229751-47229751 on Build GRCh38
rs745681472 Chr.20: 47229771-47229771 on Build GRCh38
rs999900029 Chr.20: 47229744-47229744 on Build GRCh38
rs6018350 Chr.20: 47229659-47229659 on Build GRCh38
rs773605276 Chr.20: 47229837-47229837 on Build GRCh38
rs749827144 Chr.20: 47229750-47229750 on Build GRCh38
rs776010392 Chr.20: 47229824-47229824 on Build GRCh38
rs1045581901 Chr.20: 47229739-47229739 on Build GRCh38
rs771974375 Chr.20: 47229779-47229779 on Build GRCh38
rs771358856 Chr.20: 47229847-47229847 on Build GRCh38
rs781259477 Chr.20: 47229746-47229746 on Build GRCh38
rs761488270 Chr.20: 47229826-47229826 on Build GRCh38
rs764820722 Chr.20: 47229834-47229834 on Build GRCh38
rs550264347 Chr.20: 47229794-47229794 on Build GRCh38
rs779602820 Chr.20: 47229823-47229824 on Build GRCh38
rs778727193 Chr.20: 47229765-47229765 on Build GRCh38
rs6124983 Chr.20: 47229807-47229807 on Build GRCh38
rs368217103 Chr.20: 47229678-47229678 on Build GRCh38
rs746516627 Chr.20: 47229803-47229803 on Build GRCh38
rs937088972 Chr.20: 47229764-47229764 on Build GRCh38
rs755583896 Chr.20: 47229814-47229815 on Build GRCh38
rs755179890 Chr.20: 47229734-47229734 on Build GRCh38

View Full Product Details