Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 10790692-10790963 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDIA6
Entrez Gene ID
10130
Gene Name
protein disulfide isomerase family A member 6
Gene Aliases
ERP5, P5, TXNDC7
UniGene
Hs.212102
Gene Chromosome Location
Chr.2: 10783391-10837977 on Build GRCh38
Transcripts
NM_001282704.1 NM_001282705.1 NM_001282706.1 NM_001282707.1 NM_005742.3 XM_011510308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4432199 c.647C>T p.T216M Chr.2: 10790786-10790786 on Build GRCh38
4556617 c.697G>A p.G233R Chr.2: 10790721-10790721 on Build GRCh38
714834 c.633G>A p.T211T Chr.2: 10790785-10790785 on Build GRCh38
4908938 c.660G>A p.V220V Chr.2: 10790773-10790773 on Build GRCh38
1751969 c.645G>A p.V215V Chr.2: 10790773-10790773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769286711 Chr.2: 10790812-10790812 on Build GRCh38
rs983615363 Chr.2: 10790938-10790938 on Build GRCh38
rs777563756 Chr.2: 10790886-10790886 on Build GRCh38
rs200721282 Chr.2: 10790850-10790850 on Build GRCh38
rs1039601878 Chr.2: 10790796-10790796 on Build GRCh38
rs11545887 Chr.2: 10790733-10790733 on Build GRCh38
rs750347480 Chr.2: 10790874-10790874 on Build GRCh38
rs4807 Chr.2: 10790777-10790777 on Build GRCh38
rs752689582 Chr.2: 10790762-10790762 on Build GRCh38
rs145487229 Chr.2: 10790785-10790785 on Build GRCh38
rs149768142 Chr.2: 10790744-10790744 on Build GRCh38
rs1734420 Chr.2: 10790937-10790937 on Build GRCh38
rs761212881 Chr.2: 10790811-10790811 on Build GRCh38
rs199868833 Chr.2: 10790856-10790856 on Build GRCh38
rs773600727 Chr.2: 10790821-10790821 on Build GRCh38
rs778704221 Chr.2: 10790771-10790771 on Build GRCh38
rs766666586 Chr.2: 10790743-10790743 on Build GRCh38
rs971568423 Chr.2: 10790894-10790894 on Build GRCh38
rs899685061 Chr.2: 10790799-10790799 on Build GRCh38
rs375510023 Chr.2: 10790837-10790837 on Build GRCh38
rs763479949 Chr.2: 10790826-10790826 on Build GRCh38
rs1035740110 Chr.2: 10790934-10790934 on Build GRCh38
rs752220050 Chr.2: 10790870-10790870 on Build GRCh38
rs747071044 Chr.2: 10790850-10790854 on Build GRCh38
rs759795149 Chr.2: 10790842-10790842 on Build GRCh38
rs1614107 Chr.2: 10790875-10790875 on Build GRCh38
rs775472901 Chr.2: 10790834-10790834 on Build GRCh38
rs766756529 Chr.2: 10790832-10790832 on Build GRCh38
rs755148170 Chr.2: 10790749-10790749 on Build GRCh38
rs776267394 Chr.2: 10790804-10790804 on Build GRCh38
rs781299912 Chr.2: 10790760-10790760 on Build GRCh38
rs1004658621 Chr.2: 10790849-10790849 on Build GRCh38
rs768388716 Chr.2: 10790793-10790793 on Build GRCh38
rs1734419 Chr.2: 10790920-10790920 on Build GRCh38
rs1015206814 Chr.2: 10790941-10790941 on Build GRCh38
rs370617777 Chr.2: 10790868-10790868 on Build GRCh38
rs780196239 Chr.2: 10790765-10790765 on Build GRCh38
rs35871206 Chr.2: 10790721-10790722 on Build GRCh38
rs182465865 Chr.2: 10790726-10790726 on Build GRCh38
rs369014875 Chr.2: 10790783-10790783 on Build GRCh38
rs769026085 Chr.2: 10790855-10790856 on Build GRCh38
rs372148080 Chr.2: 10790724-10790724 on Build GRCh38
rs905835374 Chr.2: 10790914-10790914 on Build GRCh38
rs55760342 Chr.2: 10790844-10790844 on Build GRCh38
rs757288641 Chr.2: 10790764-10790764 on Build GRCh38
rs150615615 Chr.2: 10790721-10790721 on Build GRCh38
rs779945564 Chr.2: 10790782-10790782 on Build GRCh38
rs765655090 Chr.2: 10790722-10790722 on Build GRCh38
rs745750029 Chr.2: 10790776-10790776 on Build GRCh38
rs752882248 Chr.2: 10790862-10790862 on Build GRCh38

View Full Product Details