Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50517296-50517531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYCBPAP
Entrez Gene ID
84073
Gene Name
MYCBP associated protein
Gene Aliases
AMAP-1, AMAP1
UniGene
Hs.398196
Gene Chromosome Location
Chr.17: 50507778-50531502 on Build GRCh38
Transcripts
NM_032133.4 XM_005257725.3 XM_005257726.2 XM_011525342.1 XM_011525343.1 XM_017025205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6147314 c.381C>A p.S127S Chr.17: 50517451-50517451 on Build GRCh38
981069 c.448C>T p.R150C Chr.17: 50517407-50517407 on Build GRCh38
706946 c.323G>T p.C108F Chr.17: 50517393-50517393 on Build GRCh38
6718910 c.382+1G>C p.? Chr.17: 50517453-50517453 on Build GRCh38
981067 c.406A>G p.K136E Chr.17: 50517365-50517365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201966035 Chr.17: 50517470-50517470 on Build GRCh38
rs748666174 Chr.17: 50517398-50517398 on Build GRCh38
rs778766687 Chr.17: 50517500-50517500 on Build GRCh38
rs574956641 Chr.17: 50517443-50517443 on Build GRCh38
rs754673853 Chr.17: 50517479-50517479 on Build GRCh38
rs144231508 Chr.17: 50517345-50517345 on Build GRCh38
rs375095253 Chr.17: 50517458-50517458 on Build GRCh38
rs200266994 Chr.17: 50517487-50517487 on Build GRCh38
rs141768814 Chr.17: 50517329-50517329 on Build GRCh38
rs781639636 Chr.17: 50517387-50517387 on Build GRCh38
rs182026801 Chr.17: 50517356-50517356 on Build GRCh38
rs759021550 Chr.17: 50517408-50517408 on Build GRCh38
rs368470723 Chr.17: 50517485-50517485 on Build GRCh38
rs765146254 Chr.17: 50517451-50517451 on Build GRCh38
rs114335869 Chr.17: 50517414-50517414 on Build GRCh38
rs756388380 Chr.17: 50517371-50517371 on Build GRCh38
rs145797399 Chr.17: 50517452-50517452 on Build GRCh38
rs1053468418 Chr.17: 50517389-50517389 on Build GRCh38
rs781556887 Chr.17: 50517507-50517507 on Build GRCh38
rs763840189 Chr.17: 50517339-50517339 on Build GRCh38
rs747192545 Chr.17: 50517369-50517369 on Build GRCh38
rs146573039 Chr.17: 50517357-50517357 on Build GRCh38
rs770267955 Chr.17: 50517403-50517403 on Build GRCh38
rs372372435 Chr.17: 50517418-50517418 on Build GRCh38
rs890018880 Chr.17: 50517421-50517421 on Build GRCh38
rs767129885 Chr.17: 50517324-50517324 on Build GRCh38
rs748578096 Chr.17: 50517508-50517508 on Build GRCh38
rs79000284 Chr.17: 50517343-50517343 on Build GRCh38
rs775039064 Chr.17: 50517415-50517415 on Build GRCh38
rs769162927 Chr.17: 50517373-50517373 on Build GRCh38
rs149804358 Chr.17: 50517423-50517423 on Build GRCh38
rs61749930 Chr.17: 50517330-50517330 on Build GRCh38
rs776345785 Chr.17: 50517429-50517429 on Build GRCh38
rs773750865 Chr.17: 50517407-50517407 on Build GRCh38

View Full Product Details