Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29994652-29994877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIRIP3
Entrez Gene ID
8479
Gene Name
HIRA interacting protein 3
Gene Aliases
-
UniGene
Hs.567370
Gene Chromosome Location
Chr.16: 29992321-29996096 on Build GRCh38
Transcripts
NM_001197323.1 NM_003609.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4690388 c.421_423delGAG p.E141delE Chr.16: 29994722-29994724 on Build GRCh38
379093 c.458C>T p.A153V Chr.16: 29994687-29994687 on Build GRCh38
471616 c.439C>T p.R147W Chr.16: 29994706-29994706 on Build GRCh38
6574229 c.394C>A p.P132T Chr.16: 29994751-29994751 on Build GRCh38
7316431 c.383A>G p.K128R Chr.16: 29994762-29994762 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747703572 Chr.16: 29994854-29994854 on Build GRCh38
rs767969100 Chr.16: 29994780-29994780 on Build GRCh38
rs756166294 Chr.16: 29994788-29994788 on Build GRCh38
rs868262583 Chr.16: 29994720-29994721 on Build GRCh38
rs771848295 Chr.16: 29994674-29994674 on Build GRCh38
rs775591825 Chr.16: 29994679-29994679 on Build GRCh38
rs538364131 Chr.16: 29994842-29994842 on Build GRCh38
rs779210410 Chr.16: 29994828-29994828 on Build GRCh38
rs776213695 Chr.16: 29994847-29994847 on Build GRCh38
rs930607887 Chr.16: 29994839-29994839 on Build GRCh38
rs759965240 Chr.16: 29994757-29994757 on Build GRCh38
rs765088333 Chr.16: 29994714-29994725 on Build GRCh38
rs139509453 Chr.16: 29994837-29994837 on Build GRCh38
rs776409016 Chr.16: 29994694-29994694 on Build GRCh38
rs755232805 Chr.16: 29994792-29994794 on Build GRCh38
rs764943905 Chr.16: 29994733-29994733 on Build GRCh38
rs746126115 Chr.16: 29994722-29994724 on Build GRCh38
rs1039364913 Chr.16: 29994691-29994691 on Build GRCh38
rs746998480 Chr.16: 29994838-29994838 on Build GRCh38
rs892237569 Chr.16: 29994746-29994746 on Build GRCh38
rs764220901 Chr.16: 29994813-29994813 on Build GRCh38
rs750673744 Chr.16: 29994687-29994687 on Build GRCh38
rs753246552 Chr.16: 29994787-29994787 on Build GRCh38
rs756169065 Chr.16: 29994833-29994833 on Build GRCh38
rs920238234 Chr.16: 29994737-29994737 on Build GRCh38
rs774517553 Chr.16: 29994752-29994752 on Build GRCh38
rs368601244 Chr.16: 29994824-29994824 on Build GRCh38
rs769278648 Chr.16: 29994855-29994855 on Build GRCh38
rs761437806 Chr.16: 29994706-29994706 on Build GRCh38
rs757355734 Chr.16: 29994826-29994826 on Build GRCh38
rs781361380 Chr.16: 29994685-29994685 on Build GRCh38
rs571185613 Chr.16: 29994729-29994729 on Build GRCh38

View Full Product Details