Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65595437-65595672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK7
Entrez Gene ID
10089
Gene Name
potassium two pore domain channel subfamily K member 7
Gene Aliases
K2p7.1, TWIK3
UniGene
Hs.175218
Gene Chromosome Location
Chr.11: 65592855-65595996 on Build GRCh38
Transcripts
NM_005714.1 NM_033347.1 NM_033348.1 NM_033455.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3451838 c.236C>T p.S79F Chr.11: 65595537-65595537 on Build GRCh38
7313559 c.175C>A p.P59T Chr.11: 65595598-65595598 on Build GRCh38
4524058 c.124G>A p.E42K Chr.11: 65595649-65595649 on Build GRCh38
4643579 c.240C>T p.T80T Chr.11: 65595533-65595533 on Build GRCh38
4601803 c.270G>C p.W90C Chr.11: 65595503-65595503 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756178334 Chr.11: 65595506-65595506 on Build GRCh38
rs377428429 Chr.11: 65595589-65595589 on Build GRCh38
rs759185389 Chr.11: 65595550-65595550 on Build GRCh38
rs201986391 Chr.11: 65595504-65595504 on Build GRCh38
rs779973894 Chr.11: 65595557-65595562 on Build GRCh38
rs754123086 Chr.11: 65595612-65595612 on Build GRCh38
rs955375911 Chr.11: 65595580-65595580 on Build GRCh38
rs781648189 Chr.11: 65595503-65595503 on Build GRCh38
rs752136574 Chr.11: 65595478-65595478 on Build GRCh38
rs780378321 Chr.11: 65595514-65595514 on Build GRCh38
rs774811778 Chr.11: 65595643-65595643 on Build GRCh38
rs1038884520 Chr.11: 65595620-65595620 on Build GRCh38
rs78680181 Chr.11: 65595592-65595592 on Build GRCh38
rs751915008 Chr.11: 65595586-65595586 on Build GRCh38
rs755107811 Chr.11: 65595614-65595614 on Build GRCh38
rs779174037 Chr.11: 65595616-65595616 on Build GRCh38
rs373233713 Chr.11: 65595548-65595548 on Build GRCh38
rs770849577 Chr.11: 65595630-65595630 on Build GRCh38
rs771045761 Chr.11: 65595542-65595542 on Build GRCh38
rs150893696 Chr.11: 65595468-65595468 on Build GRCh38
rs779929937 Chr.11: 65595604-65595604 on Build GRCh38
rs764538718 Chr.11: 65595577-65595577 on Build GRCh38
rs762470004 Chr.11: 65595563-65595563 on Build GRCh38
rs200665936 Chr.11: 65595647-65595647 on Build GRCh38
rs970594428 Chr.11: 65595458-65595458 on Build GRCh38
rs570596099 Chr.11: 65595537-65595537 on Build GRCh38
rs755838107 Chr.11: 65595494-65595495 on Build GRCh38
rs896954733 Chr.11: 65595547-65595547 on Build GRCh38
rs764296236 Chr.11: 65595474-65595474 on Build GRCh38
rs757711029 Chr.11: 65595479-65595479 on Build GRCh38
rs762130045 Chr.11: 65595587-65595587 on Build GRCh38
rs990785233 Chr.11: 65595633-65595633 on Build GRCh38
rs771907950 Chr.11: 65595529-65595529 on Build GRCh38
rs781339923 Chr.11: 65595631-65595631 on Build GRCh38
rs752289112 Chr.11: 65595470-65595470 on Build GRCh38
rs769374599 Chr.11: 65595640-65595640 on Build GRCh38
rs149982266 Chr.11: 65595627-65595627 on Build GRCh38
rs758857878 Chr.11: 65595472-65595472 on Build GRCh38
rs1016746731 Chr.11: 65595486-65595486 on Build GRCh38
rs769174857 Chr.11: 65595551-65595551 on Build GRCh38
rs550747611 Chr.11: 65595461-65595461 on Build GRCh38
rs756198654 Chr.11: 65595593-65595593 on Build GRCh38
rs762499417 Chr.11: 65595645-65595645 on Build GRCh38
rs773988339 Chr.11: 65595651-65595651 on Build GRCh38
rs746969521 Chr.11: 65595541-65595541 on Build GRCh38
rs745744437 Chr.11: 65595634-65595634 on Build GRCh38
rs1029667544 Chr.11: 65595567-65595567 on Build GRCh38
rs769678116 Chr.11: 65595512-65595512 on Build GRCh38
rs749410633 Chr.11: 65595515-65595515 on Build GRCh38
rs775224228 Chr.11: 65595560-65595560 on Build GRCh38

View Full Product Details