Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 94379999-94380266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR83
Entrez Gene ID
10888
Gene Name
G protein-coupled receptor 83
Gene Aliases
GIR, GPR72
UniGene
Hs.272385
Gene Chromosome Location
Chr.11: 94377311-94401479 on Build GRCh38
Transcripts
NM_016540.3 XM_011542559.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
690910 c.1246G>T p.V416L Chr.11: 94380175-94380175 on Build GRCh38
1357918 c.1173C>A p.G391G Chr.11: 94380248-94380248 on Build GRCh38
5594640 c.1266G>A p.M422I Chr.11: 94380155-94380155 on Build GRCh38
4414505 c.1165A>T p.N389Y Chr.11: 94380256-94380256 on Build GRCh38
4758910 c.1261A>T p.T421S Chr.11: 94380160-94380160 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748946515 Chr.11: 94380152-94380152 on Build GRCh38
rs953731927 Chr.11: 94380076-94380076 on Build GRCh38
rs766164482 Chr.11: 94380223-94380223 on Build GRCh38
rs753547935 Chr.11: 94380228-94380228 on Build GRCh38
rs267603253 Chr.11: 94380173-94380173 on Build GRCh38
rs146036633 Chr.11: 94380235-94380235 on Build GRCh38
rs199759506 Chr.11: 94380128-94380128 on Build GRCh38
rs768927714 Chr.11: 94380195-94380195 on Build GRCh38
rs565778386 Chr.11: 94380086-94380086 on Build GRCh38
rs372761443 Chr.11: 94380190-94380190 on Build GRCh38
rs144674806 Chr.11: 94380159-94380159 on Build GRCh38
rs1017030274 Chr.11: 94380224-94380224 on Build GRCh38
rs778089232 Chr.11: 94380023-94380023 on Build GRCh38
rs1053873232 Chr.11: 94380157-94380157 on Build GRCh38
rs751590202 Chr.11: 94380121-94380121 on Build GRCh38
rs888074994 Chr.11: 94380177-94380177 on Build GRCh38
rs867546572 Chr.11: 94380020-94380020 on Build GRCh38
rs973020518 Chr.11: 94380029-94380029 on Build GRCh38
rs779592376 Chr.11: 94380140-94380140 on Build GRCh38
rs1003931143 Chr.11: 94380209-94380209 on Build GRCh38
rs1049422048 Chr.11: 94380082-94380082 on Build GRCh38
rs149389877 Chr.11: 94380158-94380158 on Build GRCh38
rs747396417 Chr.11: 94380235-94380235 on Build GRCh38
rs750322440 Chr.11: 94380220-94380220 on Build GRCh38
rs528951531 Chr.11: 94380196-94380196 on Build GRCh38
rs535466654 Chr.11: 94380019-94380019 on Build GRCh38
rs757854013 Chr.11: 94380239-94380239 on Build GRCh38
rs374848600 Chr.11: 94380127-94380127 on Build GRCh38
rs369214380 Chr.11: 94380160-94380160 on Build GRCh38
rs376409703 Chr.11: 94380216-94380216 on Build GRCh38
rs3740867 Chr.11: 94380122-94380122 on Build GRCh38
rs755895172 Chr.11: 94380138-94380138 on Build GRCh38
rs781580441 Chr.11: 94380240-94380240 on Build GRCh38
rs752216621 Chr.11: 94380236-94380236 on Build GRCh38
rs774563255 Chr.11: 94380202-94380202 on Build GRCh38
rs939805997 Chr.11: 94380181-94380181 on Build GRCh38
rs547483627 Chr.11: 94380080-94380080 on Build GRCh38
rs763150214 Chr.11: 94380120-94380120 on Build GRCh38
rs754521562 Chr.11: 94380154-94380154 on Build GRCh38
rs977502522 Chr.11: 94380123-94380123 on Build GRCh38
rs775626000 Chr.11: 94380186-94380186 on Build GRCh38
rs781675419 Chr.11: 94380165-94380165 on Build GRCh38
rs148089068 Chr.11: 94380170-94380170 on Build GRCh38
rs771502713 Chr.11: 94380185-94380188 on Build GRCh38
rs745323031 Chr.11: 94380136-94380136 on Build GRCh38
rs775742094 Chr.11: 94380119-94380119 on Build GRCh38
rs747976780 Chr.11: 94380079-94380079 on Build GRCh38
rs141761818 Chr.11: 94380221-94380221 on Build GRCh38
rs530913081 Chr.11: 94380116-94380116 on Build GRCh38
rs1030562503 Chr.11: 94380042-94380042 on Build GRCh38
rs200133196 Chr.11: 94380185-94380185 on Build GRCh38

View Full Product Details