Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35939960-35940201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRFN3
Entrez Gene ID
79414
Gene Name
leucine rich repeat and fibronectin type III domain containing 3
Gene Aliases
FIGLER1, SALM4
UniGene
Hs.143792
Gene Chromosome Location
Chr.19: 35937120-35945195 on Build GRCh38
Transcripts
NM_024509.1 XM_017027302.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6431964 c.680G>C p.R227P Chr.19: 35940105-35940105 on Build GRCh38
5601749 c.602C>T p.S201F Chr.19: 35940027-35940027 on Build GRCh38
3371276 c.595G>A p.A199T Chr.19: 35940020-35940020 on Build GRCh38
6901395 c.628G>T p.D210Y Chr.19: 35940053-35940053 on Build GRCh38
1256686 c.676T>C p.S226P Chr.19: 35940101-35940101 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200648362 Chr.19: 35940101-35940101 on Build GRCh38
rs962660051 Chr.19: 35940046-35940046 on Build GRCh38
rs776754660 Chr.19: 35940047-35940047 on Build GRCh38
rs767574670 Chr.19: 35940138-35940138 on Build GRCh38
rs749851389 Chr.19: 35940140-35940140 on Build GRCh38
rs149386300 Chr.19: 35940013-35940013 on Build GRCh38
rs747770044 Chr.19: 35940137-35940137 on Build GRCh38
rs146057944 Chr.19: 35940166-35940166 on Build GRCh38
rs747501966 Chr.19: 35940105-35940105 on Build GRCh38
rs867616411 Chr.19: 35940029-35940029 on Build GRCh38
rs1006928267 Chr.19: 35940027-35940027 on Build GRCh38
rs55822021 Chr.19: 35940171-35940171 on Build GRCh38
rs555620207 Chr.19: 35940016-35940016 on Build GRCh38
rs770062400 Chr.19: 35940049-35940049 on Build GRCh38
rs781070537 Chr.19: 35940119-35940119 on Build GRCh38
rs774350545 Chr.19: 35940136-35940136 on Build GRCh38
rs201737864 Chr.19: 35940118-35940118 on Build GRCh38
rs752965843 Chr.19: 35940095-35940095 on Build GRCh38
rs556142420 Chr.19: 35940039-35940039 on Build GRCh38
rs757706780 Chr.19: 35940176-35940176 on Build GRCh38
rs765848921 Chr.19: 35940154-35940154 on Build GRCh38
rs1034795650 Chr.19: 35939995-35939995 on Build GRCh38
rs780898352 Chr.19: 35940003-35940003 on Build GRCh38
rs760265795 Chr.19: 35940145-35940145 on Build GRCh38
rs201857342 Chr.19: 35940088-35940088 on Build GRCh38
rs754683298 Chr.19: 35940161-35940161 on Build GRCh38
rs781735033 Chr.19: 35940181-35940181 on Build GRCh38
rs537848197 Chr.19: 35940036-35940036 on Build GRCh38
rs761895890 Chr.19: 35940082-35940082 on Build GRCh38
rs751516231 Chr.19: 35940069-35940069 on Build GRCh38
rs759676327 Chr.19: 35940048-35940048 on Build GRCh38
rs751929847 Chr.19: 35940169-35940169 on Build GRCh38
rs768553940 Chr.19: 35940133-35940133 on Build GRCh38
rs767131061 Chr.19: 35940085-35940085 on Build GRCh38
rs199609203 Chr.19: 35940068-35940068 on Build GRCh38
rs769862426 Chr.19: 35940128-35940128 on Build GRCh38
rs892462709 Chr.19: 35940178-35940178 on Build GRCh38
rs7247644 Chr.19: 35940100-35940100 on Build GRCh38
rs866394662 Chr.19: 35940045-35940045 on Build GRCh38
rs756795965 Chr.19: 35939999-35939999 on Build GRCh38
rs116617870 Chr.19: 35939977-35939977 on Build GRCh38
rs111616155 Chr.19: 35940149-35940149 on Build GRCh38
rs779776456 Chr.19: 35940094-35940094 on Build GRCh38
rs372082676 Chr.19: 35939986-35939986 on Build GRCh38
rs879165600 Chr.19: 35940134-35940134 on Build GRCh38
rs748558175 Chr.19: 35940019-35940019 on Build GRCh38
rs762835262 Chr.19: 35940056-35940056 on Build GRCh38
rs947800280 Chr.19: 35940132-35940132 on Build GRCh38
rs369511702 Chr.19: 35940160-35940160 on Build GRCh38
rs376226745 Chr.19: 35940129-35940129 on Build GRCh38
rs143716568 Chr.19: 35940014-35940014 on Build GRCh38
rs200697946 Chr.19: 35940030-35940030 on Build GRCh38
rs371947967 Chr.19: 35940050-35940050 on Build GRCh38
rs745976472 Chr.19: 35940127-35940127 on Build GRCh38

View Full Product Details