Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6599186-6599443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL21
Entrez Gene ID
9903
Gene Name
kelch like family member 21
Gene Aliases
-
UniGene
Hs.7764
Gene Chromosome Location
Chr.1: 6590724-6602898 on Build GRCh38
Transcripts
NM_001324309.1 NM_014851.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1743661 c.1281C>T p.I427I Chr.1: 6599193-6599193 on Build GRCh38
5468134 c.1089G>A p.A363A Chr.1: 6599385-6599385 on Build GRCh38
6006473 c.1119G>T p.E373D Chr.1: 6599355-6599355 on Build GRCh38
5484040 c.1165G>A p.A389T Chr.1: 6599309-6599309 on Build GRCh38
6696609 c.1256C>T p.A419V Chr.1: 6599218-6599218 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756603318 Chr.1: 6599258-6599258 on Build GRCh38
rs960681090 Chr.1: 6599340-6599340 on Build GRCh38
rs370117216 Chr.1: 6599343-6599343 on Build GRCh38
rs773454858 Chr.1: 6599376-6599376 on Build GRCh38
rs746528367 Chr.1: 6599292-6599292 on Build GRCh38
rs527537968 Chr.1: 6599213-6599213 on Build GRCh38
rs768198844 Chr.1: 6599318-6599319 on Build GRCh38
rs760087795 Chr.1: 6599386-6599386 on Build GRCh38
rs538024704 Chr.1: 6599306-6599306 on Build GRCh38
rs775185852 Chr.1: 6599301-6599301 on Build GRCh38
rs771634290 Chr.1: 6599212-6599212 on Build GRCh38
rs757010590 Chr.1: 6599419-6599419 on Build GRCh38
rs750944814 Chr.1: 6599319-6599320 on Build GRCh38
rs907488885 Chr.1: 6599344-6599344 on Build GRCh38
rs780894318 Chr.1: 6599346-6599346 on Build GRCh38
rs375759686 Chr.1: 6599305-6599305 on Build GRCh38
rs775339390 Chr.1: 6599360-6599360 on Build GRCh38
rs767599768 Chr.1: 6599256-6599256 on Build GRCh38
rs1052509083 Chr.1: 6599229-6599229 on Build GRCh38
rs776508686 Chr.1: 6599223-6599223 on Build GRCh38
rs368398322 Chr.1: 6599262-6599262 on Build GRCh38
rs768119966 Chr.1: 6599330-6599330 on Build GRCh38
rs372952934 Chr.1: 6599307-6599307 on Build GRCh38
rs751824361 Chr.1: 6599247-6599247 on Build GRCh38
rs762367664 Chr.1: 6599315-6599316 on Build GRCh38
rs761423081 Chr.1: 6599420-6599420 on Build GRCh38
rs937458501 Chr.1: 6599349-6599349 on Build GRCh38
rs111512176 Chr.1: 6599394-6599394 on Build GRCh38
rs780554801 Chr.1: 6599261-6599261 on Build GRCh38
rs141248147 Chr.1: 6599294-6599294 on Build GRCh38
rs769605366 Chr.1: 6599297-6599297 on Build GRCh38
rs965841298 Chr.1: 6599272-6599273 on Build GRCh38
rs753035869 Chr.1: 6599399-6599399 on Build GRCh38
rs567438879 Chr.1: 6599267-6599267 on Build GRCh38
rs367630698 Chr.1: 6599264-6599264 on Build GRCh38
rs770586174 Chr.1: 6599333-6599333 on Build GRCh38
rs766492553 Chr.1: 6599218-6599218 on Build GRCh38
rs200887044 Chr.1: 6599358-6599358 on Build GRCh38
rs764181808 Chr.1: 6599233-6599233 on Build GRCh38
rs571713856 Chr.1: 6599322-6599322 on Build GRCh38
rs753071921 Chr.1: 6599324-6599324 on Build GRCh38
rs756587903 Chr.1: 6599323-6599324 on Build GRCh38
rs139415576 Chr.1: 6599377-6599377 on Build GRCh38
rs779862641 Chr.1: 6599422-6599422 on Build GRCh38
rs143141370 Chr.1: 6599232-6599232 on Build GRCh38
rs777293501 Chr.1: 6599327-6599327 on Build GRCh38
rs199577266 Chr.1: 6599266-6599266 on Build GRCh38
rs771664109 Chr.1: 6599278-6599278 on Build GRCh38
rs762086777 Chr.1: 6599302-6599302 on Build GRCh38
rs760660606 Chr.1: 6599217-6599217 on Build GRCh38
rs550042971 Chr.1: 6599319-6599319 on Build GRCh38
rs762671661 Chr.1: 6599367-6599367 on Build GRCh38
rs200258370 Chr.1: 6599293-6599293 on Build GRCh38
rs751230069 Chr.1: 6599406-6599406 on Build GRCh38
rs777335079 Chr.1: 6599286-6599286 on Build GRCh38
rs994192011 Chr.1: 6599323-6599323 on Build GRCh38
rs754369822 Chr.1: 6599310-6599310 on Build GRCh38
rs747576093 Chr.1: 6599206-6599206 on Build GRCh38
rs772271791 Chr.1: 6599368-6599368 on Build GRCh38
rs2232461 Chr.1: 6599385-6599385 on Build GRCh38
rs979195196 Chr.1: 6599384-6599384 on Build GRCh38
rs766428949 Chr.1: 6599381-6599381 on Build GRCh38
rs554094853 Chr.1: 6599359-6599359 on Build GRCh38
rs139131803 Chr.1: 6599309-6599309 on Build GRCh38
rs756019476 Chr.1: 6599421-6599421 on Build GRCh38
rs924993186 Chr.1: 6599207-6599207 on Build GRCh38
rs757466075 Chr.1: 6599252-6599252 on Build GRCh38
rs758057941 Chr.1: 6599325-6599325 on Build GRCh38
rs143989562 Chr.1: 6599400-6599400 on Build GRCh38
rs766881707 Chr.1: 6599337-6599338 on Build GRCh38
rs2232462 Chr.1: 6599331-6599331 on Build GRCh38

View Full Product Details