Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38987787-38988042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTRA4
Entrez Gene ID
203100
Gene Name
HtrA serine peptidase 4
Gene Aliases
-
UniGene
Hs.661014
Gene Chromosome Location
Chr.8: 38974149-38988662 on Build GRCh38
Transcripts
NM_153692.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
141229 c.1361C>T p.S454F Chr.8: 38988028-38988028 on Build GRCh38
6180971 c.1310G>A p.G437E Chr.8: 38987977-38987977 on Build GRCh38
4898302 c.1287C>T p.H429H Chr.8: 38987954-38987954 on Build GRCh38
1099694 c.1282G>A p.D428N Chr.8: 38987949-38987949 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749655775 Chr.8: 38987976-38987976 on Build GRCh38
rs775728793 Chr.8: 38988006-38988006 on Build GRCh38
rs748089539 Chr.8: 38988007-38988007 on Build GRCh38
rs762632041 Chr.8: 38987933-38987933 on Build GRCh38
rs777007062 Chr.8: 38988002-38988004 on Build GRCh38
rs528808361 Chr.8: 38987868-38987868 on Build GRCh38
rs146591490 Chr.8: 38987815-38987815 on Build GRCh38
rs374401682 Chr.8: 38987956-38987956 on Build GRCh38
rs761850928 Chr.8: 38987909-38987909 on Build GRCh38
rs919078381 Chr.8: 38987970-38987970 on Build GRCh38
rs181400374 Chr.8: 38987875-38987875 on Build GRCh38
rs753144646 Chr.8: 38987959-38987959 on Build GRCh38
rs746497371 Chr.8: 38987922-38987923 on Build GRCh38
rs997066751 Chr.8: 38987816-38987816 on Build GRCh38
rs777467635 Chr.8: 38987915-38987915 on Build GRCh38
rs772215054 Chr.8: 38988004-38988004 on Build GRCh38
rs928636467 Chr.8: 38988018-38988018 on Build GRCh38
rs758083596 Chr.8: 38987914-38987915 on Build GRCh38
rs1028674922 Chr.8: 38987823-38987823 on Build GRCh38
rs1007444105 Chr.8: 38987878-38987878 on Build GRCh38
rs201501667 Chr.8: 38987931-38987931 on Build GRCh38
rs150884623 Chr.8: 38987949-38987949 on Build GRCh38
rs200076196 Chr.8: 38987907-38987907 on Build GRCh38
rs765069922 Chr.8: 38987914-38987914 on Build GRCh38
rs867434767 Chr.8: 38987941-38987941 on Build GRCh38
rs139374339 Chr.8: 38988000-38988000 on Build GRCh38
rs767017947 Chr.8: 38987957-38987957 on Build GRCh38
rs199569946 Chr.8: 38987961-38987961 on Build GRCh38
rs778195964 Chr.8: 38987975-38987975 on Build GRCh38
rs576647651 Chr.8: 38987954-38987954 on Build GRCh38
rs568701379 Chr.8: 38987876-38987876 on Build GRCh38
rs368429001 Chr.8: 38988002-38988002 on Build GRCh38
rs141552350 Chr.8: 38987936-38987936 on Build GRCh38
rs772303183 Chr.8: 38987892-38987892 on Build GRCh38
rs757481045 Chr.8: 38987989-38987989 on Build GRCh38
rs778715096 Chr.8: 38987912-38987913 on Build GRCh38
rs796928637 Chr.8: 38987912-38987913 on Build GRCh38
rs771630706 Chr.8: 38987927-38987928 on Build GRCh38

View Full Product Details