Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50245770-50246043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDAC10
Entrez Gene ID
83933
Gene Name
histone deacetylase 10
Gene Aliases
HD10
UniGene
Hs.26593
Gene Chromosome Location
Chr.22: 50245183-50251405 on Build GRCh38
Transcripts
NM_001159286.1 NM_032019.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6352464 c.1834A>G p.N612D Chr.22: 50245827-50245827 on Build GRCh38
5455056 c.1884G>T p.E628D Chr.22: 50245777-50245777 on Build GRCh38
1731816 c.1747C>A p.H583N Chr.22: 50245996-50245996 on Build GRCh38
4004962 c.1757A>G p.Q586R Chr.22: 50245986-50245986 on Build GRCh38
3842929 c.1753C>G p.L585V Chr.22: 50245990-50245990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780517680 Chr.22: 50245839-50245839 on Build GRCh38
rs867353705 Chr.22: 50245809-50245809 on Build GRCh38
rs755525016 Chr.22: 50245895-50245895 on Build GRCh38
rs902120624 Chr.22: 50245846-50245846 on Build GRCh38
rs972615726 Chr.22: 50246002-50246002 on Build GRCh38
rs764891465 Chr.22: 50245881-50245881 on Build GRCh38
rs778106529 Chr.22: 50245928-50245928 on Build GRCh38
rs753141770 Chr.22: 50245907-50245907 on Build GRCh38
rs543147576 Chr.22: 50245982-50245982 on Build GRCh38
rs368982991 Chr.22: 50245794-50245794 on Build GRCh38
rs13058054 Chr.22: 50245855-50245855 on Build GRCh38
rs753292399 Chr.22: 50245995-50245995 on Build GRCh38
rs75596977 Chr.22: 50245827-50245827 on Build GRCh38
rs953675939 Chr.22: 50245856-50245856 on Build GRCh38
rs773389854 Chr.22: 50245859-50245859 on Build GRCh38
rs868531698 Chr.22: 50245913-50245913 on Build GRCh38
rs764660372 Chr.22: 50245833-50245833 on Build GRCh38
rs751087965 Chr.22: 50245984-50245984 on Build GRCh38
rs756538758 Chr.22: 50245996-50245996 on Build GRCh38
rs771119405 Chr.22: 50245938-50245938 on Build GRCh38
rs201027749 Chr.22: 50245960-50245960 on Build GRCh38
rs764014436 Chr.22: 50245823-50245823 on Build GRCh38
rs770148642 Chr.22: 50245858-50245858 on Build GRCh38
rs762654201 Chr.22: 50245980-50245980 on Build GRCh38
rs5771271 Chr.22: 50245883-50245883 on Build GRCh38
rs755295781 Chr.22: 50245848-50245848 on Build GRCh38
rs972369098 Chr.22: 50245793-50245793 on Build GRCh38
rs866151461 Chr.22: 50245998-50245998 on Build GRCh38
rs201720171 Chr.22: 50245945-50245945 on Build GRCh38
rs917562532 Chr.22: 50245941-50245941 on Build GRCh38
rs779062288 Chr.22: 50245942-50245942 on Build GRCh38
rs781347911 Chr.22: 50245900-50245900 on Build GRCh38
rs146510222 Chr.22: 50245860-50245860 on Build GRCh38
rs953378675 Chr.22: 50245978-50245978 on Build GRCh38
rs760352851 Chr.22: 50245821-50245821 on Build GRCh38
rs758854737 Chr.22: 50245838-50245838 on Build GRCh38
rs373054489 Chr.22: 50245909-50245909 on Build GRCh38
rs776239767 Chr.22: 50245955-50245955 on Build GRCh38
rs893192076 Chr.22: 50245834-50245834 on Build GRCh38
rs13058062 Chr.22: 50245867-50245867 on Build GRCh38
rs200130118 Chr.22: 50245850-50245850 on Build GRCh38
rs778337295 Chr.22: 50246005-50246005 on Build GRCh38
rs768096692 Chr.22: 50245992-50245992 on Build GRCh38
rs763989471 Chr.22: 50245835-50245835 on Build GRCh38
rs768669155 Chr.22: 50245951-50245951 on Build GRCh38
rs772848333 Chr.22: 50245976-50245976 on Build GRCh38
rs756946870 Chr.22: 50245828-50245828 on Build GRCh38
rs749585143 Chr.22: 50245933-50245933 on Build GRCh38
rs533436968 Chr.22: 50245957-50245957 on Build GRCh38
rs11553697 Chr.22: 50245986-50245986 on Build GRCh38
rs13054930 Chr.22: 50245912-50245912 on Build GRCh38
rs761413713 Chr.22: 50245877-50245877 on Build GRCh38
rs527647798 Chr.22: 50245966-50245966 on Build GRCh38
rs775543653 Chr.22: 50245804-50245804 on Build GRCh38
rs748263087 Chr.22: 50245851-50245851 on Build GRCh38
rs769618185 Chr.22: 50245975-50245975 on Build GRCh38
rs757555958 Chr.22: 50246024-50246024 on Build GRCh38
rs768275458 Chr.22: 50245866-50245866 on Build GRCh38
rs749967021 Chr.22: 50245837-50245837 on Build GRCh38
rs751999001 Chr.22: 50245892-50245892 on Build GRCh38
rs929644724 Chr.22: 50245817-50245817 on Build GRCh38
rs760720725 Chr.22: 50245865-50245865 on Build GRCh38
rs1054475225 Chr.22: 50245829-50245829 on Build GRCh38
rs981097705 Chr.22: 50245950-50245950 on Build GRCh38
rs754052787 Chr.22: 50246010-50246010 on Build GRCh38

View Full Product Details