Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 121479576-121479809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFR2
Entrez Gene ID
2263
Gene Name
fibroblast growth factor receptor 2
Gene Aliases
BBDS, BEK, BFR-1, CD332, CEK3, CFD1, ECT1, JWS, K-SAM, KGFR, TK14, TK25
UniGene
Hs.533683
Gene Chromosome Location
Chr.10: 121478330-121598656 on Build GRCh38
Transcripts
NM_000141.4 NM_001144914.1 NM_001144915.1 NM_001144916.1 NM_001144917.1 NM_001144918.1 NM_001320654.1 NM_001320658.1 NM_022970.3 NM_023029.2 NR_073009.1 XM_006717708.2 XM_006717710.3 XM_006717711.2 XM_006717712.2 XM_017015920.1 XM_017015921.1 XM_017015922.1 XM_017015923.1 XM_017015924.1 XM_017015925.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3686533 c.2103C>T p.C701C Chr.10: 121479602-121479602 on Build GRCh38
7259178 c.2111C>A p.A704D Chr.10: 121479594-121479594 on Build GRCh38
7263516 c.2088delC p.S697fs*>11 Chr.10: 121479617-121479617 on Build GRCh38
147070 c.2107C>T p.L703L Chr.10: 121479598-121479598 on Build GRCh38
6279746 c.2035-4C>A p.? Chr.10: 121479674-121479674 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571201278 Chr.10: 121479786-121479786 on Build GRCh38
rs1047057 Chr.10: 121479598-121479598 on Build GRCh38
rs764231570 Chr.10: 121479606-121479606 on Build GRCh38
rs758066441 Chr.10: 121479716-121479716 on Build GRCh38
rs879927285 Chr.10: 121479613-121479613 on Build GRCh38
rs549203570 Chr.10: 121479674-121479674 on Build GRCh38
rs937866691 Chr.10: 121479664-121479664 on Build GRCh38
rs753575189 Chr.10: 121479684-121479684 on Build GRCh38
rs1048391923 Chr.10: 121479762-121479762 on Build GRCh38
rs748777325 Chr.10: 121479660-121479660 on Build GRCh38
rs1047721497 Chr.10: 121479752-121479752 on Build GRCh38
rs867737544 Chr.10: 121479692-121479692 on Build GRCh38
rs763095219 Chr.10: 121479601-121479601 on Build GRCh38
rs778501615 Chr.10: 121479690-121479690 on Build GRCh38
rs184026174 Chr.10: 121479707-121479707 on Build GRCh38
rs560611144 Chr.10: 121479662-121479662 on Build GRCh38
rs887507243 Chr.10: 121479770-121479770 on Build GRCh38
rs1047058 Chr.10: 121479599-121479599 on Build GRCh38
rs777204983 Chr.10: 121479720-121479720 on Build GRCh38
rs936096266 Chr.10: 121479610-121479610 on Build GRCh38
rs4647917 Chr.10: 121479673-121479673 on Build GRCh38
rs562615339 Chr.10: 121479682-121479682 on Build GRCh38
rs574474794 Chr.10: 121479746-121479746 on Build GRCh38
rs925125524 Chr.10: 121479617-121479617 on Build GRCh38
rs768166411 Chr.10: 121479705-121479706 on Build GRCh38
rs751685012 Chr.10: 121479616-121479616 on Build GRCh38

View Full Product Details