Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 41395358-41395623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MX2
Entrez Gene ID
4600
Gene Name
MX dynamin like GTPase 2
Gene Aliases
MXB
UniGene
Hs.926
Gene Chromosome Location
Chr.21: 41361986-41408943 on Build GRCh38
Transcripts
NM_002463.1 XM_005260983.4 XM_005260984.1 XM_011529571.1 XM_011529572.1 XM_011529573.1 XM_011529574.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1327393 c.894A>T p.L298L Chr.21: 41395609-41395609 on Build GRCh38
1030853 c.906C>T p.G302G Chr.21: 41395621-41395621 on Build GRCh38
6383581 c.908C>T p.T303I Chr.21: 41395623-41395623 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs973458046 Chr.21: 41395503-41395503 on Build GRCh38
rs199539451 Chr.21: 41395577-41395577 on Build GRCh38
rs187267319 Chr.21: 41395490-41395490 on Build GRCh38
rs764033139 Chr.21: 41395494-41395494 on Build GRCh38
rs752347390 Chr.21: 41395432-41395432 on Build GRCh38
rs768960271 Chr.21: 41395544-41395544 on Build GRCh38
rs766187586 Chr.21: 41395600-41395600 on Build GRCh38
rs776811501 Chr.21: 41395550-41395550 on Build GRCh38
rs528262746 Chr.21: 41395388-41395388 on Build GRCh38
rs780762872 Chr.21: 41395417-41395417 on Build GRCh38
rs751318156 Chr.21: 41395601-41395601 on Build GRCh38
rs796705966 Chr.21: 41395447-41395447 on Build GRCh38
rs982785373 Chr.21: 41395473-41395473 on Build GRCh38
rs368191593 Chr.21: 41395560-41395560 on Build GRCh38
rs76536627 Chr.21: 41395408-41395408 on Build GRCh38
rs1004288812 Chr.21: 41395450-41395450 on Build GRCh38
rs147533543 Chr.21: 41395587-41395587 on Build GRCh38
rs943939289 Chr.21: 41395573-41395574 on Build GRCh38
rs748931483 Chr.21: 41395554-41395554 on Build GRCh38
rs544403995 Chr.21: 41395421-41395421 on Build GRCh38
rs756425357 Chr.21: 41395559-41395559 on Build GRCh38
rs73370155 Chr.21: 41395486-41395486 on Build GRCh38
rs982539745 Chr.21: 41395554-41395554 on Build GRCh38
rs889818975 Chr.21: 41395513-41395513 on Build GRCh38
rs950842717 Chr.21: 41395543-41395543 on Build GRCh38
rs920671984 Chr.21: 41395528-41395528 on Build GRCh38
rs1049822320 Chr.21: 41395407-41395407 on Build GRCh38
rs769837317 Chr.21: 41395561-41395561 on Build GRCh38
rs949631326 Chr.21: 41395398-41395398 on Build GRCh38
rs761786911 Chr.21: 41395558-41395558 on Build GRCh38
rs886825021 Chr.21: 41395446-41395446 on Build GRCh38

View Full Product Details