Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 25453472-25453703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NINL
Entrez Gene ID
22981
Gene Name
ninein like
Gene Aliases
NLP
UniGene
Hs.631508
Gene Chromosome Location
Chr.20: 25452697-25585531 on Build GRCh38
Transcripts
NM_001318226.1 NM_025176.5 XM_011529186.2 XM_011529187.2 XM_011529188.1 XM_011529189.1 XM_011529190.1 XM_011529191.1 XM_011529192.1 XM_011529193.1 XM_011529194.2 XM_011529195.2 XM_011529197.2 XM_011529198.1 XM_017027732.1 XM_017027733.1 XM_017027734.1 XM_017027735.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
291222 c.3985C>T p.L1329L Chr.20: 25453615-25453615 on Build GRCh38
4097283 c.4060G>A p.A1354T Chr.20: 25453540-25453540 on Build GRCh38
6726302 c.4098C>T p.R1366R Chr.20: 25453502-25453502 on Build GRCh38
5857535 c.3977C>T p.S1326F Chr.20: 25453623-25453623 on Build GRCh38
5949043 c.3978C>A p.S1326S Chr.20: 25453622-25453622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749674928 Chr.20: 25453563-25453563 on Build GRCh38
rs772908930 Chr.20: 25453568-25453569 on Build GRCh38
rs772681395 Chr.20: 25453575-25453575 on Build GRCh38
rs765176452 Chr.20: 25453545-25453545 on Build GRCh38
rs369509557 Chr.20: 25453502-25453502 on Build GRCh38
rs776768773 Chr.20: 25453661-25453661 on Build GRCh38
rs759009567 Chr.20: 25453646-25453646 on Build GRCh38
rs140163855 Chr.20: 25453595-25453595 on Build GRCh38
rs534326565 Chr.20: 25453650-25453650 on Build GRCh38
rs760334830 Chr.20: 25453630-25453633 on Build GRCh38
rs748488492 Chr.20: 25453638-25453638 on Build GRCh38
rs761985881 Chr.20: 25453514-25453514 on Build GRCh38
rs755775647 Chr.20: 25453619-25453619 on Build GRCh38
rs146439964 Chr.20: 25453512-25453512 on Build GRCh38
rs779413806 Chr.20: 25453568-25453568 on Build GRCh38
rs531568222 Chr.20: 25453574-25453574 on Build GRCh38
rs776524973 Chr.20: 25453538-25453538 on Build GRCh38
rs547606450 Chr.20: 25453667-25453667 on Build GRCh38
rs757849636 Chr.20: 25453498-25453498 on Build GRCh38
rs762686893 Chr.20: 25453678-25453678 on Build GRCh38
rs769227074 Chr.20: 25453510-25453510 on Build GRCh38
rs562856131 Chr.20: 25453559-25453559 on Build GRCh38
rs544300410 Chr.20: 25453668-25453669 on Build GRCh38
rs6132834 Chr.20: 25453607-25453607 on Build GRCh38
rs367640910 Chr.20: 25453589-25453589 on Build GRCh38
rs144950952 Chr.20: 25453583-25453583 on Build GRCh38
rs773408351 Chr.20: 25453520-25453520 on Build GRCh38
rs752565890 Chr.20: 25453546-25453546 on Build GRCh38
rs532624786 Chr.20: 25453525-25453525 on Build GRCh38
rs772342007 Chr.20: 25453517-25453517 on Build GRCh38
rs372633499 Chr.20: 25453680-25453680 on Build GRCh38
rs751048368 Chr.20: 25453553-25453553 on Build GRCh38
rs200080729 Chr.20: 25453540-25453540 on Build GRCh38
rs369121474 Chr.20: 25453671-25453671 on Build GRCh38
rs34802534 Chr.20: 25453527-25453527 on Build GRCh38
rs747128057 Chr.20: 25453656-25453656 on Build GRCh38
rs755230297 Chr.20: 25453567-25453567 on Build GRCh38
rs758329949 Chr.20: 25453547-25453547 on Build GRCh38
rs6115189 Chr.20: 25453618-25453618 on Build GRCh38
rs746135868 Chr.20: 25453663-25453663 on Build GRCh38
rs1050112330 Chr.20: 25453524-25453524 on Build GRCh38
rs372159498 Chr.20: 25453651-25453651 on Build GRCh38
rs771219915 Chr.20: 25453651-25453667 on Build GRCh38
rs147951975 Chr.20: 25453585-25453585 on Build GRCh38
rs769981180 Chr.20: 25453664-25453664 on Build GRCh38
rs756661899 Chr.20: 25453602-25453602 on Build GRCh38
rs768594932 Chr.20: 25453679-25453679 on Build GRCh38
rs370353125 Chr.20: 25453594-25453594 on Build GRCh38
rs17857107 Chr.20: 25453503-25453503 on Build GRCh38
rs770852705 Chr.20: 25453659-25453659 on Build GRCh38
rs548178520 Chr.20: 25453597-25453597 on Build GRCh38
rs200042917 Chr.20: 25453598-25453598 on Build GRCh38
rs779464685 Chr.20: 25453622-25453622 on Build GRCh38
rs767202930 Chr.20: 25453611-25453611 on Build GRCh38
rs776663077 Chr.20: 25453587-25453587 on Build GRCh38

View Full Product Details