Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 22843220-22843481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP14
Entrez Gene ID
4323
Gene Name
matrix metallopeptidase 14
Gene Aliases
MMP-14, MMP-X1, MT-MMP, MT-MMP 1, MT1-MMP, MT1MMP, MTMMP1, WNCHRS
UniGene
Hs.2399
Gene Chromosome Location
Chr.14: 22836533-22847600 on Build GRCh38
Transcripts
NM_004995.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2030571 c.833G>A p.G278D Chr.14: 22843401-22843401 on Build GRCh38
4577761 c.741C>T p.L247L Chr.14: 22843309-22843309 on Build GRCh38
416241 c.754G>T p.D252Y Chr.14: 22843322-22843322 on Build GRCh38
6251973 c.810G>T p.V270V Chr.14: 22843378-22843378 on Build GRCh38
4572544 c.779T>A p.F260Y Chr.14: 22843347-22843347 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757917193 Chr.14: 22843426-22843426 on Build GRCh38
rs140757040 Chr.14: 22843438-22843438 on Build GRCh38
rs142651327 Chr.14: 22843398-22843398 on Build GRCh38
rs745755221 Chr.14: 22843424-22843424 on Build GRCh38
rs1021104196 Chr.14: 22843379-22843379 on Build GRCh38
rs959005393 Chr.14: 22843444-22843444 on Build GRCh38
rs765715939 Chr.14: 22843386-22843386 on Build GRCh38
rs375991979 Chr.14: 22843366-22843366 on Build GRCh38
rs201684823 Chr.14: 22843337-22843337 on Build GRCh38
rs943286775 Chr.14: 22843357-22843357 on Build GRCh38
rs369766994 Chr.14: 22843433-22843433 on Build GRCh38
rs764316658 Chr.14: 22843243-22843243 on Build GRCh38
rs911878094 Chr.14: 22843355-22843355 on Build GRCh38
rs374632825 Chr.14: 22843365-22843365 on Build GRCh38
rs771529027 Chr.14: 22843338-22843338 on Build GRCh38
rs562455694 Chr.14: 22843250-22843250 on Build GRCh38
rs757044765 Chr.14: 22843420-22843420 on Build GRCh38
rs200987693 Chr.14: 22843448-22843448 on Build GRCh38
rs769069897 Chr.14: 22843382-22843382 on Build GRCh38
rs920288322 Chr.14: 22843402-22843402 on Build GRCh38
rs200519097 Chr.14: 22843280-22843280 on Build GRCh38
rs766751794 Chr.14: 22843281-22843281 on Build GRCh38
rs894051901 Chr.14: 22843294-22843294 on Build GRCh38
rs372126942 Chr.14: 22843334-22843334 on Build GRCh38
rs776927679 Chr.14: 22843384-22843384 on Build GRCh38
rs370892055 Chr.14: 22843267-22843267 on Build GRCh38
rs755999999 Chr.14: 22843289-22843289 on Build GRCh38
rs767669970 Chr.14: 22843458-22843458 on Build GRCh38
rs753622411 Chr.14: 22843412-22843412 on Build GRCh38
rs773882324 Chr.14: 22843437-22843437 on Build GRCh38
rs762161209 Chr.14: 22843264-22843264 on Build GRCh38
rs1042704 Chr.14: 22843385-22843385 on Build GRCh38
rs150962356 Chr.14: 22843309-22843309 on Build GRCh38
rs145239346 Chr.14: 22843394-22843394 on Build GRCh38
rs746743029 Chr.14: 22843395-22843395 on Build GRCh38
rs779472403 Chr.14: 22843428-22843428 on Build GRCh38
rs778828438 Chr.14: 22843330-22843330 on Build GRCh38
rs2236302 Chr.14: 22843345-22843345 on Build GRCh38
rs17884841 Chr.14: 22843265-22843265 on Build GRCh38
rs751831406 Chr.14: 22843396-22843396 on Build GRCh38
rs137906578 Chr.14: 22843397-22843397 on Build GRCh38
rs752923075 Chr.14: 22843459-22843459 on Build GRCh38
rs548260059 Chr.14: 22843285-22843285 on Build GRCh38
rs376251631 Chr.14: 22843434-22843434 on Build GRCh38
rs143061668 Chr.14: 22843310-22843310 on Build GRCh38
rs757197165 Chr.14: 22843327-22843327 on Build GRCh38
rs539736479 Chr.14: 22843431-22843431 on Build GRCh38
rs778745322 Chr.14: 22843421-22843421 on Build GRCh38

View Full Product Details