Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 56989674-56989899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR5AK2
Entrez Gene ID
390181
Gene Name
olfactory receptor family 5 subfamily AK member 2
Gene Aliases
-
UniGene
Hs.554520
Gene Chromosome Location
Chr.11: 56988914-56989843 on Build GRCh38
Transcripts
NM_001005323.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
364984 c.769T>A p.Y257N Chr.11: 56989682-56989682 on Build GRCh38
5952698 c.821C>T p.A274V Chr.11: 56989734-56989734 on Build GRCh38
353751 c.770A>G p.Y257C Chr.11: 56989683-56989683 on Build GRCh38
6237911 c.824T>C p.F275S Chr.11: 56989737-56989737 on Build GRCh38
233004 c.805G>A p.E269K Chr.11: 56989718-56989718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759801348 Chr.11: 56989776-56989776 on Build GRCh38
rs775517468 Chr.11: 56989863-56989863 on Build GRCh38
rs573692967 Chr.11: 56989848-56989848 on Build GRCh38
rs887199590 Chr.11: 56989724-56989724 on Build GRCh38
rs767837203 Chr.11: 56989850-56989850 on Build GRCh38
rs770395238 Chr.11: 56989741-56989741 on Build GRCh38
rs116682075 Chr.11: 56989830-56989830 on Build GRCh38
rs764102363 Chr.11: 56989791-56989791 on Build GRCh38
rs371870476 Chr.11: 56989821-56989821 on Build GRCh38
rs767694663 Chr.11: 56989777-56989777 on Build GRCh38
rs774634554 Chr.11: 56989868-56989868 on Build GRCh38
rs907387035 Chr.11: 56989825-56989825 on Build GRCh38
rs745474873 Chr.11: 56989725-56989725 on Build GRCh38
rs865807027 Chr.11: 56989758-56989758 on Build GRCh38
rs758193924 Chr.11: 56989815-56989815 on Build GRCh38
rs773535349 Chr.11: 56989742-56989742 on Build GRCh38
rs773801487 Chr.11: 56989826-56989826 on Build GRCh38
rs1046804494 Chr.11: 56989770-56989770 on Build GRCh38
rs551465149 Chr.11: 56989773-56989773 on Build GRCh38
rs923177231 Chr.11: 56989704-56989704 on Build GRCh38
rs776617385 Chr.11: 56989812-56989812 on Build GRCh38
rs755847790 Chr.11: 56989786-56989786 on Build GRCh38
rs375044294 Chr.11: 56989837-56989837 on Build GRCh38
rs748461163 Chr.11: 56989737-56989737 on Build GRCh38
rs1051095938 Chr.11: 56989715-56989715 on Build GRCh38
rs777096396 Chr.11: 56989733-56989733 on Build GRCh38
rs146542991 Chr.11: 56989708-56989708 on Build GRCh38
rs190872743 Chr.11: 56989820-56989820 on Build GRCh38
rs745342220 Chr.11: 56989804-56989804 on Build GRCh38
rs756855620 Chr.11: 56989802-56989802 on Build GRCh38
rs749316678 Chr.11: 56989768-56989768 on Build GRCh38
rs753647939 Chr.11: 56989799-56989799 on Build GRCh38
rs142316901 Chr.11: 56989778-56989778 on Build GRCh38
rs774580162 Chr.11: 56989774-56989774 on Build GRCh38
rs771830861 Chr.11: 56989730-56989730 on Build GRCh38
rs1001711455 Chr.11: 56989829-56989829 on Build GRCh38
rs778709287 Chr.11: 56989717-56989717 on Build GRCh38
rs140086630 Chr.11: 56989763-56989763 on Build GRCh38
rs775607731 Chr.11: 56989747-56989747 on Build GRCh38

View Full Product Details