Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157134021-157134292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETV3
Entrez Gene ID
2117
Gene Name
ETS variant 3
Gene Aliases
METS, PE-1, PE1, bA110J1.4
UniGene
Hs.105636
Gene Chromosome Location
Chr.1: 157121191-157138591 on Build GRCh38
Transcripts
NM_001145312.1 NM_005240.1 XM_006711210.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3976251 c.318A>C p.T106T Chr.1: 157134194-157134194 on Build GRCh38
5844025 c.367C>T p.P123S Chr.1: 157134145-157134145 on Build GRCh38
5745727 c.400+8A>G p.? Chr.1: 157134104-157134104 on Build GRCh38
4395866 c.313A>G p.K105E Chr.1: 157134199-157134199 on Build GRCh38
1127340 c.290A>G p.Y97C Chr.1: 157134222-157134222 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764404209 Chr.1: 157134185-157134185 on Build GRCh38
rs772147809 Chr.1: 157134097-157134097 on Build GRCh38
rs980425502 Chr.1: 157134141-157134141 on Build GRCh38
rs746960246 Chr.1: 157134119-157134119 on Build GRCh38
rs377561731 Chr.1: 157134200-157134200 on Build GRCh38
rs758048645 Chr.1: 157134244-157134244 on Build GRCh38
rs778189711 Chr.1: 157134060-157134060 on Build GRCh38
rs201917206 Chr.1: 157134096-157134096 on Build GRCh38
rs778593027 Chr.1: 157134071-157134071 on Build GRCh38
rs200671237 Chr.1: 157134237-157134237 on Build GRCh38
rs35463779 Chr.1: 157134243-157134244 on Build GRCh38
rs200533472 Chr.1: 157134246-157134246 on Build GRCh38
rs139827367 Chr.1: 157134158-157134158 on Build GRCh38
rs775501800 Chr.1: 157134102-157134102 on Build GRCh38
rs377498585 Chr.1: 157134159-157134159 on Build GRCh38
rs745773653 Chr.1: 157134256-157134256 on Build GRCh38
rs142863577 Chr.1: 157134121-157134121 on Build GRCh38
rs756878385 Chr.1: 157134196-157134196 on Build GRCh38
rs201404958 Chr.1: 157134247-157134247 on Build GRCh38
rs766539831 Chr.1: 157134130-157134130 on Build GRCh38
rs761226318 Chr.1: 157134160-157134160 on Build GRCh38
rs752640825 Chr.1: 157134090-157134090 on Build GRCh38
rs371631974 Chr.1: 157134248-157134248 on Build GRCh38
rs201364337 Chr.1: 157134150-157134150 on Build GRCh38
rs754188589 Chr.1: 157134195-157134195 on Build GRCh38
rs35010936 Chr.1: 157134124-157134124 on Build GRCh38
rs768391921 Chr.1: 157134120-157134120 on Build GRCh38
rs748939441 Chr.1: 157134062-157134062 on Build GRCh38
rs943987500 Chr.1: 157134125-157134125 on Build GRCh38
rs888666050 Chr.1: 157134065-157134065 on Build GRCh38
rs780874277 Chr.1: 157134255-157134255 on Build GRCh38
rs756900055 Chr.1: 157134063-157134063 on Build GRCh38
rs759549727 Chr.1: 157134155-157134155 on Build GRCh38
rs778647895 Chr.1: 157134218-157134218 on Build GRCh38
rs749957817 Chr.1: 157134226-157134226 on Build GRCh38
rs932304528 Chr.1: 157134221-157134221 on Build GRCh38

View Full Product Details