Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 121993212-121993483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ILDR1
Entrez Gene ID
286676
Gene Name
immunoglobulin like domain containing receptor 1
Gene Aliases
DFNB42, ILDR1alpha, ILDR1alpha', ILDR1beta
UniGene
Hs.98484
Gene Chromosome Location
Chr.3: 121987323-122060554 on Build GRCh38
Transcripts
NM_001199799.1 NM_001199800.1 NM_175924.3 XM_005247389.4 XM_011512738.2 XM_011512739.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3586114 c.1247G>A p.G416E Chr.3: 121993370-121993370 on Build GRCh38
3940234 c.1220G>A p.S407N Chr.3: 121993397-121993397 on Build GRCh38
4318275 c.1216C>T p.P406S Chr.3: 121993437-121993437 on Build GRCh38
1693297 c.1370C>T p.S457F Chr.3: 121993247-121993247 on Build GRCh38
5344322 c.1134C>T p.S378S Chr.3: 121993483-121993483 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745614639 Chr.3: 121993242-121993242 on Build GRCh38
rs756048732 Chr.3: 121993399-121993399 on Build GRCh38
rs140567004 Chr.3: 121993452-121993452 on Build GRCh38
rs747247492 Chr.3: 121993413-121993413 on Build GRCh38
rs760020579 Chr.3: 121993310-121993310 on Build GRCh38
rs774472194 Chr.3: 121993247-121993247 on Build GRCh38
rs760818014 Chr.3: 121993294-121993294 on Build GRCh38
rs749946454 Chr.3: 121993397-121993397 on Build GRCh38
rs759967990 Chr.3: 121993392-121993392 on Build GRCh38
rs751115626 Chr.3: 121993328-121993328 on Build GRCh38
rs745810698 Chr.3: 121993350-121993350 on Build GRCh38
rs866906195 Chr.3: 121993230-121993230 on Build GRCh38
rs189004426 Chr.3: 121993288-121993288 on Build GRCh38
rs912861659 Chr.3: 121993246-121993246 on Build GRCh38
rs727503097 Chr.3: 121993389-121993389 on Build GRCh38
rs760017535 Chr.3: 121993370-121993370 on Build GRCh38
rs764388719 Chr.3: 121993390-121993390 on Build GRCh38
rs758111234 Chr.3: 121993322-121993322 on Build GRCh38
rs367565247 Chr.3: 121993258-121993258 on Build GRCh38
rs533076598 Chr.3: 121993410-121993410 on Build GRCh38
rs376986803 Chr.3: 121993392-121993392 on Build GRCh38
rs771171871 Chr.3: 121993415-121993415 on Build GRCh38
rs748441415 Chr.3: 121993284-121993284 on Build GRCh38
rs773467432 Chr.3: 121993289-121993289 on Build GRCh38
rs201639358 Chr.3: 121993249-121993249 on Build GRCh38
rs149188951 Chr.3: 121993271-121993271 on Build GRCh38
rs775062249 Chr.3: 121993365-121993365 on Build GRCh38
rs1033754995 Chr.3: 121993318-121993318 on Build GRCh38
rs753781354 Chr.3: 121993406-121993406 on Build GRCh38
rs200630651 Chr.3: 121993364-121993364 on Build GRCh38
rs760257421 Chr.3: 121993448-121993448 on Build GRCh38
rs777035964 Chr.3: 121993303-121993303 on Build GRCh38
rs781193912 Chr.3: 121993419-121993419 on Build GRCh38
rs180838731 Chr.3: 121993407-121993407 on Build GRCh38
rs144649620 Chr.3: 121993252-121993252 on Build GRCh38
rs769876173 Chr.3: 121993361-121993362 on Build GRCh38
rs774922988 Chr.3: 121993391-121993392 on Build GRCh38
rs34883204 Chr.3: 121993424-121993424 on Build GRCh38
rs769485101 Chr.3: 121993273-121993273 on Build GRCh38
rs794727852 Chr.3: 121993457-121993457 on Build GRCh38
rs544136698 Chr.3: 121993300-121993300 on Build GRCh38
rs1042728259 Chr.3: 121993405-121993405 on Build GRCh38
rs372952499 Chr.3: 121993327-121993327 on Build GRCh38
rs1004936666 Chr.3: 121993312-121993312 on Build GRCh38
rs201105694 Chr.3: 121993372-121993372 on Build GRCh38
rs370978987 Chr.3: 121993352-121993352 on Build GRCh38
rs748215825 Chr.3: 121993260-121993260 on Build GRCh38
rs372564314 Chr.3: 121993391-121993391 on Build GRCh38
rs771335457 Chr.3: 121993264-121993264 on Build GRCh38
rs146620043 Chr.3: 121993259-121993259 on Build GRCh38
rs770839837 Chr.3: 121993238-121993238 on Build GRCh38
rs371286984 Chr.3: 121993260-121993260 on Build GRCh38
rs747787444 Chr.3: 121993232-121993232 on Build GRCh38
rs138277824 Chr.3: 121993447-121993447 on Build GRCh38
rs746514835 Chr.3: 121993265-121993265 on Build GRCh38
rs778163752 Chr.3: 121993362-121993362 on Build GRCh38
rs559855953 Chr.3: 121993371-121993371 on Build GRCh38
rs756068339 Chr.3: 121993351-121993351 on Build GRCh38
rs749062569 Chr.3: 121993355-121993355 on Build GRCh38
rs781450878 Chr.3: 121993257-121993258 on Build GRCh38
rs727503096 Chr.3: 121993316-121993316 on Build GRCh38
rs767126013 Chr.3: 121993424-121993425 on Build GRCh38
rs766963403 Chr.3: 121993257-121993257 on Build GRCh38
rs200077106 Chr.3: 121993233-121993233 on Build GRCh38
rs554966703 Chr.3: 121993437-121993437 on Build GRCh38
rs34284625 Chr.3: 121993251-121993251 on Build GRCh38
rs529264421 Chr.3: 121993349-121993349 on Build GRCh38
rs141559449 Chr.3: 121993425-121993425 on Build GRCh38
rs761401095 Chr.3: 121993396-121993396 on Build GRCh38
rs763407271 Chr.3: 121993426-121993426 on Build GRCh38
rs749753650 Chr.3: 121993363-121993364 on Build GRCh38
rs35906279 Chr.3: 121993451-121993451 on Build GRCh38
rs753090807 Chr.3: 121993320-121993320 on Build GRCh38
rs895948062 Chr.3: 121993435-121993435 on Build GRCh38
rs369175544 Chr.3: 121993338-121993338 on Build GRCh38
rs35597690 Chr.3: 121993444-121993444 on Build GRCh38
rs780019730 Chr.3: 121993401-121993401 on Build GRCh38
rs771428050 Chr.3: 121993369-121993369 on Build GRCh38
rs777321422 Chr.3: 121993263-121993263 on Build GRCh38
rs556882881 Chr.3: 121993272-121993272 on Build GRCh38
rs775302314 Chr.3: 121993276-121993276 on Build GRCh38
rs774688440 Chr.3: 121993439-121993439 on Build GRCh38
rs778369601 Chr.3: 121993245-121993245 on Build GRCh38
rs757986659 Chr.3: 121993262-121993262 on Build GRCh38
rs199571441 Chr.3: 121993361-121993361 on Build GRCh38
rs146413045 Chr.3: 121993241-121993241 on Build GRCh38
rs757224204 Chr.3: 121993268-121993268 on Build GRCh38
rs755248446 Chr.3: 121993244-121993244 on Build GRCh38
rs761333724 Chr.3: 121993253-121993253 on Build GRCh38
rs376510686 Chr.3: 121993458-121993458 on Build GRCh38
rs746959964 Chr.3: 121993234-121993234 on Build GRCh38
rs756778818 Chr.3: 121993348-121993348 on Build GRCh38
rs369062793 Chr.3: 121993306-121993306 on Build GRCh38
rs148350512 Chr.3: 121993423-121993423 on Build GRCh38

View Full Product Details