Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141150366-141150637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHB6
Entrez Gene ID
56130
Gene Name
protocadherin beta 6
Gene Aliases
PCDH-BETA6
UniGene
Hs.283085
Gene Chromosome Location
Chr.5: 141150022-141153287 on Build GRCh38
Transcripts
NM_001303145.1 NM_018939.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5567038 c.271C>T p.R91W Chr.5: 141150528-141150528 on Build GRCh38
3242291 c.361C>T p.R121* Chr.5: 141150618-141150618 on Build GRCh38
7382906 c.151C>A p.L51M Chr.5: 141150408-141150408 on Build GRCh38
1062616 c.356C>T p.S119F Chr.5: 141150613-141150613 on Build GRCh38
5867572 c.148G>A p.D50N Chr.5: 141150405-141150405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782192777 Chr.5: 141150510-141150510 on Build GRCh38
rs782020814 Chr.5: 141150414-141150414 on Build GRCh38
rs548482487 Chr.5: 141150460-141150460 on Build GRCh38
rs781940602 Chr.5: 141150438-141150438 on Build GRCh38
rs782065715 Chr.5: 141150458-141150458 on Build GRCh38
rs370624837 Chr.5: 141150445-141150445 on Build GRCh38
rs782499788 Chr.5: 141150486-141150486 on Build GRCh38
rs782297197 Chr.5: 141150432-141150432 on Build GRCh38
rs782343115 Chr.5: 141150527-141150527 on Build GRCh38
rs782313925 Chr.5: 141150433-141150433 on Build GRCh38
rs782815865 Chr.5: 141150575-141150575 on Build GRCh38
rs200056604 Chr.5: 141150463-141150463 on Build GRCh38
rs1021826769 Chr.5: 141150496-141150496 on Build GRCh38
rs1000901518 Chr.5: 141150578-141150578 on Build GRCh38
rs782656665 Chr.5: 141150498-141150498 on Build GRCh38
rs782257197 Chr.5: 141150439-141150439 on Build GRCh38
rs891979677 Chr.5: 141150444-141150444 on Build GRCh38
rs781870580 Chr.5: 141150566-141150566 on Build GRCh38
rs781947621 Chr.5: 141150557-141150557 on Build GRCh38
rs782033966 Chr.5: 141150549-141150549 on Build GRCh38
rs782473233 Chr.5: 141150598-141150598 on Build GRCh38
rs146165668 Chr.5: 141150551-141150551 on Build GRCh38
rs781991742 Chr.5: 141150441-141150441 on Build GRCh38
rs781797638 Chr.5: 141150586-141150586 on Build GRCh38
rs140181050 Chr.5: 141150556-141150556 on Build GRCh38
rs1002964750 Chr.5: 141150418-141150418 on Build GRCh38
rs566877700 Chr.5: 141150544-141150544 on Build GRCh38
rs782612248 Chr.5: 141150529-141150529 on Build GRCh38
rs782146801 Chr.5: 141150442-141150442 on Build GRCh38
rs782252027 Chr.5: 141150533-141150533 on Build GRCh38
rs782720073 Chr.5: 141150538-141150538 on Build GRCh38
rs782585896 Chr.5: 141150501-141150501 on Build GRCh38
rs200997921 Chr.5: 141150574-141150574 on Build GRCh38
rs781896993 Chr.5: 141150499-141150499 on Build GRCh38
rs781789488 Chr.5: 141150457-141150457 on Build GRCh38
rs782120835 Chr.5: 141150404-141150404 on Build GRCh38
rs969039332 Chr.5: 141150528-141150528 on Build GRCh38
rs782155972 Chr.5: 141150567-141150567 on Build GRCh38
rs782558809 Chr.5: 141150500-141150500 on Build GRCh38

View Full Product Details