Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44740824-44741093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF2C
Entrez Gene ID
11004
Gene Name
kinesin family member 2C
Gene Aliases
CT139, KNSL6, MCAK
UniGene
Hs.720061
Gene Chromosome Location
Chr.1: 44739704-44767767 on Build GRCh38
Transcripts
NM_001297655.1 NM_006845.3 XM_011540540.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6318820 c.148G>T p.A50S Chr.1: 44740990-44740990 on Build GRCh38
5497541 c.75G>A p.V25V Chr.1: 44740944-44740944 on Build GRCh38
910039 c.163G>T p.E55* Chr.1: 44741005-44741005 on Build GRCh38
3985097 c.137C>A p.A46E Chr.1: 44740979-44740979 on Build GRCh38
3985098 c.110C>A p.A37E Chr.1: 44740979-44740979 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753758030 Chr.1: 44741044-44741044 on Build GRCh38
rs1047552389 Chr.1: 44740883-44740883 on Build GRCh38
rs370557781 Chr.1: 44741057-44741057 on Build GRCh38
rs750069771 Chr.1: 44741048-44741048 on Build GRCh38
rs747239443 Chr.1: 44740850-44740850 on Build GRCh38
rs995112010 Chr.1: 44740872-44740872 on Build GRCh38
rs184038569 Chr.1: 44741066-44741066 on Build GRCh38
rs749629249 Chr.1: 44740979-44740979 on Build GRCh38
rs552334552 Chr.1: 44740854-44740854 on Build GRCh38
rs765072139 Chr.1: 44741047-44741047 on Build GRCh38
rs759896566 Chr.1: 44741011-44741011 on Build GRCh38
rs746501157 Chr.1: 44740908-44740908 on Build GRCh38
rs145948180 Chr.1: 44740926-44740926 on Build GRCh38
rs757974667 Chr.1: 44741056-44741056 on Build GRCh38
rs750123116 Chr.1: 44740879-44740879 on Build GRCh38
rs772276308 Chr.1: 44741012-44741012 on Build GRCh38
rs775574160 Chr.1: 44741016-44741016 on Build GRCh38
rs779603206 Chr.1: 44740906-44740906 on Build GRCh38
rs766175275 Chr.1: 44740933-44740934 on Build GRCh38
rs774578977 Chr.1: 44741054-44741054 on Build GRCh38
rs761951358 Chr.1: 44740987-44740987 on Build GRCh38
rs548341128 Chr.1: 44741045-44741045 on Build GRCh38
rs757033705 Chr.1: 44740867-44740867 on Build GRCh38
rs976962823 Chr.1: 44741043-44741043 on Build GRCh38
rs770274652 Chr.1: 44740910-44740910 on Build GRCh38
rs1001995102 Chr.1: 44740890-44740890 on Build GRCh38
rs978843828 Chr.1: 44741063-44741064 on Build GRCh38
rs776899338 Chr.1: 44740871-44740871 on Build GRCh38
rs376774709 Chr.1: 44740874-44740874 on Build GRCh38
rs748909105 Chr.1: 44741035-44741035 on Build GRCh38
rs375173098 Chr.1: 44741004-44741004 on Build GRCh38
rs1008801574 Chr.1: 44740928-44740928 on Build GRCh38
rs779692682 Chr.1: 44740931-44740931 on Build GRCh38
rs375349838 Chr.1: 44740932-44740932 on Build GRCh38
rs996763279 Chr.1: 44741027-44741027 on Build GRCh38
rs142016314 Chr.1: 44740982-44740982 on Build GRCh38
rs762555549 Chr.1: 44740881-44740882 on Build GRCh38
rs149444880 Chr.1: 44741029-44741029 on Build GRCh38
rs189702486 Chr.1: 44740894-44740894 on Build GRCh38
rs768986926 Chr.1: 44740857-44740857 on Build GRCh38
rs368007780 Chr.1: 44741053-44741053 on Build GRCh38

View Full Product Details