Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65006028-65006267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTFMT
Entrez Gene ID
123263
Gene Name
mitochondrial methionyl-tRNA formyltransferase
Gene Aliases
COXPD15, FMT1
UniGene
Hs.531615
Gene Chromosome Location
Chr.15: 65001512-65029639 on Build GRCh38
Transcripts
NM_139242.3 XM_005254158.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2256204 c.828G>A p.T276T Chr.15: 65006177-65006177 on Build GRCh38
1215651 c.851A>C p.K284T Chr.15: 65006154-65006154 on Build GRCh38
2256206 c.827C>T p.T276M Chr.15: 65006178-65006178 on Build GRCh38
3503129 c.881C>T p.S294L Chr.15: 65006124-65006124 on Build GRCh38
5485751 c.818C>T p.P273L Chr.15: 65006187-65006187 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs906317671 Chr.15: 65006090-65006090 on Build GRCh38
rs587777418 Chr.15: 65006127-65006127 on Build GRCh38
rs369374908 Chr.15: 65006073-65006073 on Build GRCh38
rs753491878 Chr.15: 65006165-65006165 on Build GRCh38
rs755719151 Chr.15: 65006070-65006070 on Build GRCh38
rs202082622 Chr.15: 65006120-65006120 on Build GRCh38
rs762770982 Chr.15: 65006118-65006118 on Build GRCh38
rs769828080 Chr.15: 65006223-65006223 on Build GRCh38
rs758795979 Chr.15: 65006081-65006081 on Build GRCh38
rs780530646 Chr.15: 65006177-65006177 on Build GRCh38
rs755424500 Chr.15: 65006181-65006181 on Build GRCh38
rs747614238 Chr.15: 65006180-65006180 on Build GRCh38
rs530492914 Chr.15: 65006096-65006096 on Build GRCh38
rs752434860 Chr.15: 65006067-65006067 on Build GRCh38
rs377758917 Chr.15: 65006160-65006160 on Build GRCh38
rs776567068 Chr.15: 65006099-65006099 on Build GRCh38
rs370239519 Chr.15: 65006188-65006188 on Build GRCh38
rs752137723 Chr.15: 65006149-65006149 on Build GRCh38
rs758882234 Chr.15: 65006166-65006166 on Build GRCh38
rs575133242 Chr.15: 65006148-65006148 on Build GRCh38
rs777996180 Chr.15: 65006235-65006235 on Build GRCh38
rs774396833 Chr.15: 65006123-65006123 on Build GRCh38
rs114097513 Chr.15: 65006158-65006158 on Build GRCh38
rs747447473 Chr.15: 65006089-65006089 on Build GRCh38
rs768816503 Chr.15: 65006094-65006094 on Build GRCh38
rs777508477 Chr.15: 65006071-65006071 on Build GRCh38
rs373413541 Chr.15: 65006088-65006088 on Build GRCh38
rs748458161 Chr.15: 65006101-65006101 on Build GRCh38
rs918733303 Chr.15: 65006129-65006129 on Build GRCh38
rs773358847 Chr.15: 65006105-65006105 on Build GRCh38
rs781679969 Chr.15: 65006186-65006186 on Build GRCh38
rs770019763 Chr.15: 65006104-65006104 on Build GRCh38

View Full Product Details