Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 115966031-115966290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED13L
Entrez Gene ID
23389
Gene Name
mediator complex subunit 13 like
Gene Aliases
MRFACD, PROSIT240, THRAP2, TRAP240L
UniGene
Hs.603766
Gene Chromosome Location
Chr.12: 115958576-116277219 on Build GRCh38
Transcripts
NM_015335.4 XM_011538080.2 XM_011538081.2 XM_011538082.2 XM_017019090.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6477801 c.6306A>T p.S2102S Chr.12: 115966163-115966163 on Build GRCh38
5903022 c.6347C>T p.S2116L Chr.12: 115966122-115966122 on Build GRCh38
6310649 c.6257C>A p.A2086E Chr.12: 115966212-115966212 on Build GRCh38
3792222 c.6319G>C p.E2107Q Chr.12: 115966150-115966150 on Build GRCh38
6326730 c.6387+5G>A p.? Chr.12: 115966077-115966077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019178380 Chr.12: 115966159-115966159 on Build GRCh38
rs778789057 Chr.12: 115966111-115966111 on Build GRCh38
rs774432060 Chr.12: 115966122-115966122 on Build GRCh38
rs764199219 Chr.12: 115966259-115966259 on Build GRCh38
rs768108181 Chr.12: 115966147-115966147 on Build GRCh38
rs775809961 Chr.12: 115966227-115966227 on Build GRCh38
rs965077828 Chr.12: 115966236-115966236 on Build GRCh38
rs769874135 Chr.12: 115966121-115966121 on Build GRCh38
rs747734369 Chr.12: 115966168-115966168 on Build GRCh38
rs760001110 Chr.12: 115966210-115966210 on Build GRCh38
rs768045639 Chr.12: 115966220-115966220 on Build GRCh38
rs760837237 Chr.12: 115966252-115966252 on Build GRCh38
rs764431907 Chr.12: 115966062-115966062 on Build GRCh38
rs754070933 Chr.12: 115966067-115966067 on Build GRCh38
rs868718640 Chr.12: 115966143-115966143 on Build GRCh38
rs369934604 Chr.12: 115966059-115966059 on Build GRCh38
rs1037425194 Chr.12: 115966144-115966144 on Build GRCh38
rs897429320 Chr.12: 115966169-115966169 on Build GRCh38
rs3088260 Chr.12: 115966115-115966115 on Build GRCh38
rs140340239 Chr.12: 115966190-115966190 on Build GRCh38
rs140872502 Chr.12: 115966146-115966146 on Build GRCh38
rs886041624 Chr.12: 115966184-115966185 on Build GRCh38
rs756339110 Chr.12: 115966058-115966058 on Build GRCh38
rs776298494 Chr.12: 115966148-115966148 on Build GRCh38
rs571633592 Chr.12: 115966075-115966075 on Build GRCh38

View Full Product Details