Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49889918-49890183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL4I1
Entrez Gene ID
259307
Gene Name
interleukin 4 induced 1
Gene Aliases
FIG1, LAAO, LAO
UniGene
Hs.574492
Gene Chromosome Location
Chr.19: 49889654-49929539 on Build GRCh38
Transcripts
NM_001258017.1 NM_001258018.1 NM_152899.1 NM_172374.2 NR_047577.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6376633 c.1388T>C p.V463A Chr.19: 49890052-49890052 on Build GRCh38
6378380 c.1292G>A p.G431D Chr.19: 49890148-49890148 on Build GRCh38
7268106 c.1346A>G p.H449R Chr.19: 49890028-49890028 on Build GRCh38
6574033 c.1377G>C p.P459P Chr.19: 49889997-49889997 on Build GRCh38
7268105 c.1412A>G p.H471R Chr.19: 49890028-49890028 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866921301 Chr.19: 49890091-49890091 on Build GRCh38
rs372496464 Chr.19: 49889951-49889951 on Build GRCh38
rs967141731 Chr.19: 49890002-49890002 on Build GRCh38
rs142348169 Chr.19: 49889988-49889988 on Build GRCh38
rs752017974 Chr.19: 49890129-49890129 on Build GRCh38
rs369174992 Chr.19: 49889934-49889934 on Build GRCh38
rs922207875 Chr.19: 49890070-49890070 on Build GRCh38
rs528564762 Chr.19: 49890128-49890128 on Build GRCh38
rs199648353 Chr.19: 49890105-49890105 on Build GRCh38
rs1019937807 Chr.19: 49890166-49890166 on Build GRCh38
rs757552751 Chr.19: 49890162-49890162 on Build GRCh38
rs781407372 Chr.19: 49889949-49889949 on Build GRCh38
rs574930395 Chr.19: 49890076-49890076 on Build GRCh38
rs554391826 Chr.19: 49890134-49890134 on Build GRCh38
rs541635369 Chr.19: 49889981-49889981 on Build GRCh38
rs745773961 Chr.19: 49890030-49890030 on Build GRCh38
rs774978694 Chr.19: 49890096-49890096 on Build GRCh38
rs111587296 Chr.19: 49889937-49889937 on Build GRCh38
rs933599980 Chr.19: 49890084-49890084 on Build GRCh38
rs372789046 Chr.19: 49890148-49890148 on Build GRCh38
rs1012762903 Chr.19: 49889959-49889959 on Build GRCh38
rs940783527 Chr.19: 49890093-49890093 on Build GRCh38
rs898965412 Chr.19: 49890125-49890125 on Build GRCh38
rs756340364 Chr.19: 49889952-49889952 on Build GRCh38
rs973038388 Chr.19: 49890072-49890072 on Build GRCh38
rs112829612 Chr.19: 49890149-49890149 on Build GRCh38
rs753980117 Chr.19: 49890150-49890150 on Build GRCh38
rs868229227 Chr.19: 49889950-49889950 on Build GRCh38
rs777339667 Chr.19: 49890120-49890120 on Build GRCh38
rs974930487 Chr.19: 49890053-49890053 on Build GRCh38
rs772058191 Chr.19: 49890090-49890090 on Build GRCh38
rs752768926 Chr.19: 49890140-49890140 on Build GRCh38
rs200284411 Chr.19: 49890028-49890028 on Build GRCh38
rs1051760085 Chr.19: 49890088-49890088 on Build GRCh38
rs745745610 Chr.19: 49890165-49890165 on Build GRCh38
rs761563577 Chr.19: 49890122-49890122 on Build GRCh38
rs760189496 Chr.19: 49890113-49890113 on Build GRCh38
rs1027343388 Chr.19: 49890092-49890092 on Build GRCh38
rs56359967 Chr.19: 49890012-49890012 on Build GRCh38
rs1020393732 Chr.19: 49889994-49889994 on Build GRCh38
rs978950469 Chr.19: 49890003-49890003 on Build GRCh38
rs768245215 Chr.19: 49890117-49890117 on Build GRCh38
rs751978658 Chr.19: 49889936-49889936 on Build GRCh38
rs976949046 Chr.19: 49890151-49890151 on Build GRCh38

View Full Product Details