Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10353377-10353604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TYK2
Entrez Gene ID
7297
Gene Name
tyrosine kinase 2
Gene Aliases
IMD35, JTK1
UniGene
Hs.75516
Gene Chromosome Location
Chr.19: 10350528-10380572 on Build GRCh38
Transcripts
NM_003331.4 XM_011528245.1 XM_011528246.2 XM_011528247.1 XM_011528249.2 XM_011528251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1231474 c.2974C>T p.R992W Chr.19: 10353581-10353581 on Build GRCh38
1390086 c.2996A>G p.Q999R Chr.19: 10353559-10353559 on Build GRCh38
4876359 c.2997G>A p.Q999Q Chr.19: 10353558-10353558 on Build GRCh38
6890415 c.2973C>T p.P991P Chr.19: 10353582-10353582 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370401636 Chr.19: 10353581-10353581 on Build GRCh38
rs962977869 Chr.19: 10353426-10353426 on Build GRCh38
rs758470420 Chr.19: 10353582-10353582 on Build GRCh38
rs544133815 Chr.19: 10353451-10353451 on Build GRCh38
rs573280434 Chr.19: 10353424-10353424 on Build GRCh38
rs749667172 Chr.19: 10353534-10353534 on Build GRCh38
rs914581858 Chr.19: 10353512-10353512 on Build GRCh38
rs990154876 Chr.19: 10353494-10353494 on Build GRCh38
rs992173568 Chr.19: 10353433-10353433 on Build GRCh38
rs148484862 Chr.19: 10353576-10353576 on Build GRCh38
rs551288067 Chr.19: 10353474-10353474 on Build GRCh38
rs545844060 Chr.19: 10353464-10353464 on Build GRCh38
rs147442318 Chr.19: 10353570-10353570 on Build GRCh38
rs753617141 Chr.19: 10353574-10353574 on Build GRCh38
rs371686810 Chr.19: 10353454-10353454 on Build GRCh38
rs202110875 Chr.19: 10353544-10353544 on Build GRCh38
rs1039399074 Chr.19: 10353551-10353551 on Build GRCh38
rs934634930 Chr.19: 10353492-10353492 on Build GRCh38
rs771379400 Chr.19: 10353536-10353536 on Build GRCh38
rs934914210 Chr.19: 10353448-10353448 on Build GRCh38
rs924717510 Chr.19: 10353491-10353491 on Build GRCh38
rs901645819 Chr.19: 10353562-10353562 on Build GRCh38
rs772177678 Chr.19: 10353569-10353569 on Build GRCh38
rs866757379 Chr.19: 10353537-10353537 on Build GRCh38
rs1023129667 Chr.19: 10353435-10353435 on Build GRCh38
rs764232789 Chr.19: 10353572-10353572 on Build GRCh38
rs201397594 Chr.19: 10353578-10353578 on Build GRCh38
rs865983705 Chr.19: 10353531-10353531 on Build GRCh38
rs778134416 Chr.19: 10353516-10353516 on Build GRCh38
rs768261169 Chr.19: 10353499-10353499 on Build GRCh38
rs746663364 Chr.19: 10353498-10353498 on Build GRCh38
rs561392136 Chr.19: 10353396-10353396 on Build GRCh38
rs760882131 Chr.19: 10353571-10353571 on Build GRCh38
rs760083588 Chr.19: 10353546-10353546 on Build GRCh38
rs12720359 Chr.19: 10353462-10353462 on Build GRCh38
rs1006410018 Chr.19: 10353584-10353584 on Build GRCh38

View Full Product Details