Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 94467790-94468053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCD3
Entrez Gene ID
5825
Gene Name
ATP binding cassette subfamily D member 3
Gene Aliases
ABC43, CBAS5, PMP70, PXMP1, ZWS2
UniGene
Hs.700576
Gene Chromosome Location
Chr.1: 94385106-94518663 on Build GRCh38
Transcripts
NM_001122674.1 NM_002858.3 XM_006710802.2 XM_017001952.1 XM_017001953.1 XM_017001954.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3977996 c.335G>A p.S112N Chr.1: 94468007-94468007 on Build GRCh38
3806175 c.251G>A p.G84D Chr.1: 94467923-94467923 on Build GRCh38
3493567 c.309G>A p.M103I Chr.1: 94467981-94467981 on Build GRCh38
6064489 c.324A>T p.T108T Chr.1: 94467996-94467996 on Build GRCh38
3806176 c.251G>A p.G84D Chr.1: 94467923-94467923 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs939962868 Chr.1: 94467830-94467830 on Build GRCh38
rs11558249 Chr.1: 94467965-94467965 on Build GRCh38
rs771739876 Chr.1: 94467956-94467956 on Build GRCh38
rs748806095 Chr.1: 94467945-94467945 on Build GRCh38
rs370087408 Chr.1: 94468021-94468021 on Build GRCh38
rs561652938 Chr.1: 94467885-94467885 on Build GRCh38
rs763473057 Chr.1: 94467910-94467910 on Build GRCh38
rs779274457 Chr.1: 94468011-94468012 on Build GRCh38
rs770994734 Chr.1: 94468019-94468019 on Build GRCh38
rs541543507 Chr.1: 94467934-94467934 on Build GRCh38
rs765518807 Chr.1: 94467985-94467985 on Build GRCh38
rs375038988 Chr.1: 94467972-94467972 on Build GRCh38
rs757684877 Chr.1: 94467916-94467916 on Build GRCh38
rs201160279 Chr.1: 94467899-94467899 on Build GRCh38
rs761794235 Chr.1: 94467954-94467954 on Build GRCh38
rs1036903825 Chr.1: 94467856-94467856 on Build GRCh38
rs11558250 Chr.1: 94467979-94467979 on Build GRCh38
rs955886111 Chr.1: 94468015-94468015 on Build GRCh38
rs746881972 Chr.1: 94468011-94468011 on Build GRCh38
rs768509467 Chr.1: 94467947-94467947 on Build GRCh38
rs979715405 Chr.1: 94468013-94468013 on Build GRCh38
rs371274977 Chr.1: 94467901-94467901 on Build GRCh38
rs139039035 Chr.1: 94467966-94467966 on Build GRCh38
rs773098054 Chr.1: 94467961-94467961 on Build GRCh38
rs905583862 Chr.1: 94467948-94467948 on Build GRCh38
rs774263843 Chr.1: 94467952-94467952 on Build GRCh38
rs766725060 Chr.1: 94467936-94467936 on Build GRCh38
rs1014571901 Chr.1: 94467943-94467943 on Build GRCh38
rs779387280 Chr.1: 94467942-94467942 on Build GRCh38
rs538153726 Chr.1: 94467862-94467862 on Build GRCh38
rs1035385612 Chr.1: 94468012-94468012 on Build GRCh38
rs1035294801 Chr.1: 94467969-94467979 on Build GRCh38
rs781659062 Chr.1: 94467928-94467928 on Build GRCh38

View Full Product Details