Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186076881-186077112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLR3
Entrez Gene ID
7098
Gene Name
toll like receptor 3
Gene Aliases
CD283, IIAE2
UniGene
Hs.657724
Gene Chromosome Location
Chr.4: 186069155-186086724 on Build GRCh38
Transcripts
NM_003265.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1229398 c.266A>C p.K89T Chr.4: 186076885-186076885 on Build GRCh38
7393940 c.335C>G p.S112C Chr.4: 186076954-186076954 on Build GRCh38
4491685 c.384C>T p.L128L Chr.4: 186077003-186077003 on Build GRCh38
7393937 c.275C>A p.P92Q Chr.4: 186076894-186076894 on Build GRCh38
1724344 c.288G>T p.Q96H Chr.4: 186076907-186076907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150769655 Chr.4: 186076917-186076917 on Build GRCh38
rs745694858 Chr.4: 186076952-186076952 on Build GRCh38
rs753086454 Chr.4: 186077041-186077042 on Build GRCh38
rs777194678 Chr.4: 186077051-186077051 on Build GRCh38
rs770856238 Chr.4: 186076944-186076944 on Build GRCh38
rs368416421 Chr.4: 186077074-186077074 on Build GRCh38
rs767025064 Chr.4: 186077001-186077001 on Build GRCh38
rs763487767 Chr.4: 186076988-186076988 on Build GRCh38
rs375996512 Chr.4: 186076979-186076979 on Build GRCh38
rs755773063 Chr.4: 186077045-186077045 on Build GRCh38
rs372734410 Chr.4: 186076920-186076920 on Build GRCh38
rs767814924 Chr.4: 186077029-186077029 on Build GRCh38
rs202236935 Chr.4: 186076987-186076987 on Build GRCh38
rs201726068 Chr.4: 186077006-186077006 on Build GRCh38
rs756648006 Chr.4: 186077062-186077062 on Build GRCh38
rs754548817 Chr.4: 186076914-186076914 on Build GRCh38
rs140176180 Chr.4: 186076965-186076965 on Build GRCh38
rs769267414 Chr.4: 186076935-186076935 on Build GRCh38
rs749161017 Chr.4: 186076942-186076942 on Build GRCh38
rs762007597 Chr.4: 186076972-186076972 on Build GRCh38
rs752393845 Chr.4: 186077040-186077040 on Build GRCh38
rs186586295 Chr.4: 186076945-186076945 on Build GRCh38
rs778886455 Chr.4: 186077069-186077069 on Build GRCh38
rs777335713 Chr.4: 186076939-186076939 on Build GRCh38
rs143625567 Chr.4: 186077063-186077063 on Build GRCh38
rs559783546 Chr.4: 186076957-186076957 on Build GRCh38
rs774089808 Chr.4: 186076947-186076947 on Build GRCh38
rs139173043 Chr.4: 186076928-186076928 on Build GRCh38
rs748824571 Chr.4: 186077056-186077056 on Build GRCh38
rs751822847 Chr.4: 186077005-186077005 on Build GRCh38

View Full Product Details