Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 17185056-17185311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D5
Entrez Gene ID
9779
Gene Name
TBC1 domain family member 5
Gene Aliases
-
UniGene
Hs.475629
Gene Chromosome Location
Chr.3: 17157162-17742739 on Build GRCh38
Transcripts
NM_001134381.1 NM_014744.2 XM_005265611.1 XM_005265612.1 XM_005265614.1 XM_005265615.1 XM_005265616.4 XM_006713430.1 XM_011534281.1 XM_011534283.2 XM_011534284.2 XM_011534286.1 XM_011534287.1 XM_017007552.1 XM_017007553.1 XM_017007554.1 XM_017007555.1 XM_017007556.1 XM_017007557.1 XM_017007558.1 XM_017007559.1 XM_017007560.1 XM_017007561.1 XM_017007562.1 XM_017007563.1 XM_017007564.1 XM_017007565.1 XM_017007566.1 XM_017007567.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1041344 c.1835T>C p.V612A Chr.3: 17185126-17185126 on Build GRCh38
4473411 c.1782C>T p.F594F Chr.3: 17185179-17185179 on Build GRCh38
3590286 c.1778C>T p.S593F Chr.3: 17185183-17185183 on Build GRCh38
6797476 c.1852G>C p.V618L Chr.3: 17185109-17185109 on Build GRCh38
5355386 c.1783C>A p.L595I Chr.3: 17185178-17185178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777162757 Chr.3: 17185162-17185162 on Build GRCh38
rs751345477 Chr.3: 17185236-17185236 on Build GRCh38
rs748115441 Chr.3: 17185127-17185127 on Build GRCh38
rs775138315 Chr.3: 17185146-17185146 on Build GRCh38
rs993770410 Chr.3: 17185228-17185229 on Build GRCh38
rs755574728 Chr.3: 17185145-17185145 on Build GRCh38
rs959457018 Chr.3: 17185268-17185268 on Build GRCh38
rs762088528 Chr.3: 17185227-17185227 on Build GRCh38
rs376723873 Chr.3: 17185228-17185228 on Build GRCh38
rs756406014 Chr.3: 17185236-17185237 on Build GRCh38
rs764209216 Chr.3: 17185219-17185219 on Build GRCh38
rs141235471 Chr.3: 17185174-17185174 on Build GRCh38
rs748907712 Chr.3: 17185167-17185167 on Build GRCh38
rs866613522 Chr.3: 17185183-17185183 on Build GRCh38
rs754039556 Chr.3: 17185112-17185112 on Build GRCh38
rs766129657 Chr.3: 17185117-17185117 on Build GRCh38
rs559090779 Chr.3: 17185119-17185119 on Build GRCh38
rs965583645 Chr.3: 17185252-17185252 on Build GRCh38
rs989073375 Chr.3: 17185277-17185277 on Build GRCh38
rs774768605 Chr.3: 17185188-17185188 on Build GRCh38
rs754728266 Chr.3: 17185237-17185237 on Build GRCh38
rs149215404 Chr.3: 17185109-17185109 on Build GRCh38
rs199795099 Chr.3: 17185200-17185200 on Build GRCh38
rs760873507 Chr.3: 17185085-17185085 on Build GRCh38
rs913377832 Chr.3: 17185279-17185279 on Build GRCh38
rs746270314 Chr.3: 17185191-17185191 on Build GRCh38
rs1033835350 Chr.3: 17185265-17185265 on Build GRCh38
rs754764435 Chr.3: 17185113-17185113 on Build GRCh38
rs752576095 Chr.3: 17185258-17185258 on Build GRCh38
rs963195835 Chr.3: 17185115-17185115 on Build GRCh38
rs777842638 Chr.3: 17185253-17185256 on Build GRCh38
rs754895843 Chr.3: 17185122-17185122 on Build GRCh38
rs772673188 Chr.3: 17185193-17185193 on Build GRCh38
rs776577379 Chr.3: 17185221-17185221 on Build GRCh38
rs751376239 Chr.3: 17185118-17185118 on Build GRCh38
rs781155636 Chr.3: 17185124-17185124 on Build GRCh38
rs143365571 Chr.3: 17185114-17185114 on Build GRCh38
rs994977268 Chr.3: 17185147-17185147 on Build GRCh38
rs377411512 Chr.3: 17185248-17185248 on Build GRCh38
rs1051528529 Chr.3: 17185106-17185106 on Build GRCh38
rs776096582 Chr.3: 17185196-17185196 on Build GRCh38
rs1037391768 Chr.3: 17185150-17185150 on Build GRCh38
rs374087252 Chr.3: 17185084-17185084 on Build GRCh38

View Full Product Details