Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 146218960-146219189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM27
Entrez Gene ID
54439
Gene Name
RNA binding motif protein 27
Gene Aliases
ARRS1, Psc1, ZC3H18, ZC3H20
UniGene
Hs.61441
Gene Chromosome Location
Chr.5: 146203550-146289223 on Build GRCh38
Transcripts
NM_018989.1 XM_005268466.1 XM_005268467.1 XM_005268468.1 XM_006714787.1 XM_011537661.1 XM_011537662.1 XM_017009587.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4945651 c.60-5A>G p.? Chr.5: 146218980-146218980 on Build GRCh38
3210139 c.154C>A p.Q52K Chr.5: 146219079-146219079 on Build GRCh38
4653904 c.157C>T p.L53F Chr.5: 146219082-146219082 on Build GRCh38
2151116 c.70G>A p.D24N Chr.5: 146218995-146218995 on Build GRCh38
7118806 c.60-4_67delCTAGATGTGATG p.? Chr.5: 146218981-146218992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749490733 Chr.5: 146219094-146219094 on Build GRCh38
rs537812760 Chr.5: 146219069-146219069 on Build GRCh38
rs778047273 Chr.5: 146219087-146219087 on Build GRCh38
rs771014375 Chr.5: 146219116-146219116 on Build GRCh38
rs975606372 Chr.5: 146219146-146219146 on Build GRCh38
rs764708996 Chr.5: 146219131-146219131 on Build GRCh38
rs751831006 Chr.5: 146219018-146219018 on Build GRCh38
rs774934489 Chr.5: 146219041-146219041 on Build GRCh38
rs768505055 Chr.5: 146219124-146219124 on Build GRCh38
rs371894593 Chr.5: 146219031-146219031 on Build GRCh38
rs748729581 Chr.5: 146219049-146219049 on Build GRCh38
rs755393174 Chr.5: 146219021-146219021 on Build GRCh38
rs761471569 Chr.5: 146219108-146219108 on Build GRCh38
rs982960599 Chr.5: 146219099-146219099 on Build GRCh38
rs774652067 Chr.5: 146219120-146219120 on Build GRCh38
rs965157082 Chr.5: 146219122-146219122 on Build GRCh38
rs762556579 Chr.5: 146219134-146219134 on Build GRCh38
rs376195920 Chr.5: 146219127-146219127 on Build GRCh38
rs776548411 Chr.5: 146219126-146219126 on Build GRCh38
rs369219568 Chr.5: 146219133-146219133 on Build GRCh38
rs780435614 Chr.5: 146219015-146219015 on Build GRCh38

View Full Product Details