Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 228408191-228408418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM17
Entrez Gene ID
51127
Gene Name
tripartite motif containing 17
Gene Aliases
RBCC, RNF16, terf
UniGene
Hs.121748
Gene Chromosome Location
Chr.1: 228407478-228416882 on Build GRCh38
Transcripts
NM_001024940.2 NM_016102.3 XM_006711779.2 XM_011544209.2 XM_011544210.2 XM_011544211.2 XM_017001418.1 XM_017001419.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4464604 c.1341C>T p.T447T Chr.1: 228408294-228408294 on Build GRCh38
4524345 c.1266G>A p.L422L Chr.1: 228408369-228408369 on Build GRCh38
5936952 c.1376G>A p.G459E Chr.1: 228408259-228408259 on Build GRCh38
6528123 c.1291T>G p.F431V Chr.1: 228408344-228408344 on Build GRCh38
7137570 c.1272C>T p.F424F Chr.1: 228408363-228408363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201303816 Chr.1: 228408329-228408329 on Build GRCh38
rs113469102 Chr.1: 228408264-228408264 on Build GRCh38
rs762388887 Chr.1: 228408304-228408304 on Build GRCh38
rs753823811 Chr.1: 228408258-228408258 on Build GRCh38
rs563004573 Chr.1: 228408341-228408343 on Build GRCh38
rs747294361 Chr.1: 228408285-228408285 on Build GRCh38
rs147270097 Chr.1: 228408382-228408382 on Build GRCh38
rs528401423 Chr.1: 228408330-228408330 on Build GRCh38
rs758504303 Chr.1: 228408357-228408357 on Build GRCh38
rs746074695 Chr.1: 228408269-228408269 on Build GRCh38
rs371626210 Chr.1: 228408302-228408302 on Build GRCh38
rs578077271 Chr.1: 228408355-228408355 on Build GRCh38
rs748591927 Chr.1: 228408380-228408380 on Build GRCh38
rs1025997353 Chr.1: 228408349-228408349 on Build GRCh38
rs149011573 Chr.1: 228408336-228408336 on Build GRCh38
rs200718874 Chr.1: 228408282-228408282 on Build GRCh38
rs373630367 Chr.1: 228408348-228408348 on Build GRCh38
rs146463638 Chr.1: 228408267-228408267 on Build GRCh38
rs367802352 Chr.1: 228408350-228408350 on Build GRCh38
rs371037607 Chr.1: 228408225-228408225 on Build GRCh38
rs769533359 Chr.1: 228408284-228408296 on Build GRCh38
rs767498281 Chr.1: 228408294-228408297 on Build GRCh38
rs747580337 Chr.1: 228408237-228408237 on Build GRCh38
rs61737802 Chr.1: 228408252-228408252 on Build GRCh38
rs145111971 Chr.1: 228408335-228408335 on Build GRCh38
rs769088235 Chr.1: 228408291-228408291 on Build GRCh38
rs148081432 Chr.1: 228408351-228408351 on Build GRCh38
rs531298821 Chr.1: 228408257-228408257 on Build GRCh38
rs374063435 Chr.1: 228408356-228408356 on Build GRCh38
rs142038911 Chr.1: 228408261-228408261 on Build GRCh38
rs148358732 Chr.1: 228408363-228408363 on Build GRCh38
rs772327862 Chr.1: 228408279-228408279 on Build GRCh38
rs373698198 Chr.1: 228408254-228408254 on Build GRCh38
rs770563409 Chr.1: 228408311-228408311 on Build GRCh38
rs113940247 Chr.1: 228408249-228408249 on Build GRCh38
rs370345983 Chr.1: 228408353-228408353 on Build GRCh38
rs768837283 Chr.1: 228408365-228408365 on Build GRCh38
rs780090533 Chr.1: 228408362-228408362 on Build GRCh38
rs781670355 Chr.1: 228408370-228408370 on Build GRCh38

View Full Product Details