Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 77610811-77611036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPN12
Entrez Gene ID
5782
Gene Name
protein tyrosine phosphatase, non-receptor type 12
Gene Aliases
PTP-PEST, PTPG1
UniGene
Hs.61812
Gene Chromosome Location
Chr.7: 77537160-77640071 on Build GRCh38
Transcripts
NM_001131008.1 NM_001131009.1 NM_002835.3 XM_005250518.2 XM_006716073.3 XM_011516444.1 XM_017012474.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6758820 c.884delA p.K296fs*18 Chr.7: 77610991-77610991 on Build GRCh38
6481478 c.894A>G p.L298L Chr.7: 77611001-77611001 on Build GRCh38
234078 c.841-9C>T p.? Chr.7: 77610939-77610939 on Build GRCh38
6758808 c.830T>C p.V277A Chr.7: 77610832-77610832 on Build GRCh38
4881964 c.919C>G p.Q307E Chr.7: 77611026-77611026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768368685 Chr.7: 77610905-77610907 on Build GRCh38
rs762332146 Chr.7: 77610931-77610931 on Build GRCh38
rs766487217 Chr.7: 77610836-77610836 on Build GRCh38
rs745781361 Chr.7: 77610849-77610852 on Build GRCh38
rs769667278 Chr.7: 77610849-77610861 on Build GRCh38
rs202223509 Chr.7: 77610875-77610875 on Build GRCh38
rs770081224 Chr.7: 77610889-77610891 on Build GRCh38
rs773388710 Chr.7: 77610946-77610946 on Build GRCh38
rs776923140 Chr.7: 77610922-77610922 on Build GRCh38
rs752597201 Chr.7: 77610839-77610839 on Build GRCh38
rs879214126 Chr.7: 77610886-77610886 on Build GRCh38
rs897355729 Chr.7: 77610953-77610953 on Build GRCh38
rs771809696 Chr.7: 77610884-77610884 on Build GRCh38
rs751835358 Chr.7: 77610975-77610975 on Build GRCh38
rs924126205 Chr.7: 77610887-77610887 on Build GRCh38
rs778696281 Chr.7: 77611001-77611001 on Build GRCh38
rs775113641 Chr.7: 77610894-77610894 on Build GRCh38
rs569459328 Chr.7: 77610983-77610983 on Build GRCh38
rs370891737 Chr.7: 77610989-77610989 on Build GRCh38
rs777641154 Chr.7: 77610861-77610861 on Build GRCh38
rs376113683 Chr.7: 77610897-77610897 on Build GRCh38
rs750011474 Chr.7: 77611007-77611007 on Build GRCh38
rs749247291 Chr.7: 77610870-77610870 on Build GRCh38
rs769013979 Chr.7: 77610915-77610915 on Build GRCh38
rs757262071 Chr.7: 77611000-77611000 on Build GRCh38
rs757090913 Chr.7: 77610874-77610874 on Build GRCh38
rs762927585 Chr.7: 77610956-77610956 on Build GRCh38
rs370969702 Chr.7: 77610882-77610882 on Build GRCh38
rs779570309 Chr.7: 77610870-77610872 on Build GRCh38
rs765624152 Chr.7: 77610939-77610939 on Build GRCh38
rs978633127 Chr.7: 77610873-77610873 on Build GRCh38
rs766689991 Chr.7: 77610973-77610973 on Build GRCh38
rs753644682 Chr.7: 77610998-77610998 on Build GRCh38
rs189190032 Chr.7: 77610849-77610849 on Build GRCh38

View Full Product Details