Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 128910650-128910921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAD9
Entrez Gene ID
28976
Gene Name
acyl-CoA dehydrogenase family member 9
Gene Aliases
NPD002
UniGene
Hs.567482
Gene Chromosome Location
Chr.3: 128879490-128916075 on Build GRCh38
Transcripts
NM_014049.4 NR_033426.1 XM_017006264.1 XM_017006265.1 XM_011513050.2 XM_017006939.1 XM_017006940.1 XM_017006941.1 XM_017006942.1 XM_017006943.1 XM_017006944.1 XM_017006945.1 XM_017006946.1 XM_017006947.1 XM_017006948.1 XM_017006949.1 XM_017006950.1 XM_017006951.1 XM_017006952.1 XM_017006953.1 XM_017006954.1

Gene Symbol
KIAA1257
Entrez Gene ID
57501
Gene Name
KIAA1257
Gene Aliases
-
UniGene
Hs.518247
Gene Chromosome Location
Chr.3: 128910186-129032431 on Build GRCh38
Transcripts
NM_014049.4 NR_033426.1 XM_017006264.1 XM_017006265.1 XM_011513050.2 XM_017006939.1 XM_017006940.1 XM_017006941.1 XM_017006942.1 XM_017006943.1 XM_017006944.1 XM_017006945.1 XM_017006946.1 XM_017006947.1 XM_017006948.1 XM_017006949.1 XM_017006950.1 XM_017006951.1 XM_017006952.1 XM_017006953.1 XM_017006954.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
727579 c.1719G>T p.V573V Chr.3: 128910767-128910767 on Build GRCh38
3767235 c.1716C>T p.C572C Chr.3: 128910764-128910764 on Build GRCh38
7076245 c.1743C>T p.F581F Chr.3: 128910791-128910791 on Build GRCh38
4658706 c.1736A>C p.N579T Chr.3: 128910784-128910784 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745820197 Chr.3: 128910734-128910734 on Build GRCh38
rs767137316 Chr.3: 128910773-128910773 on Build GRCh38
rs909857752 Chr.3: 128910686-128910686 on Build GRCh38
rs775299800 Chr.3: 128910754-128910754 on Build GRCh38
rs768175073 Chr.3: 128910759-128910759 on Build GRCh38
rs190759235 Chr.3: 128910891-128910891 on Build GRCh38
rs904198884 Chr.3: 128910883-128910883 on Build GRCh38
rs199572038 Chr.3: 128910843-128910843 on Build GRCh38
rs1013079776 Chr.3: 128910857-128910857 on Build GRCh38
rs779008004 Chr.3: 128910713-128910714 on Build GRCh38
rs778095534 Chr.3: 128910713-128910713 on Build GRCh38
rs183779245 Chr.3: 128910848-128910848 on Build GRCh38
rs1013062093 Chr.3: 128910833-128910833 on Build GRCh38
rs535800944 Chr.3: 128910673-128910673 on Build GRCh38
rs147040198 Chr.3: 128910764-128910764 on Build GRCh38
rs1038995992 Chr.3: 128910723-128910723 on Build GRCh38
rs191563945 Chr.3: 128910729-128910729 on Build GRCh38
rs750123246 Chr.3: 128910767-128910767 on Build GRCh38
rs756996267 Chr.3: 128910730-128910730 on Build GRCh38
rs757427510 Chr.3: 128910731-128910731 on Build GRCh38
rs758578580 Chr.3: 128910690-128910690 on Build GRCh38
rs765657785 Chr.3: 128910812-128910812 on Build GRCh38
rs773805385 Chr.3: 128910865-128910865 on Build GRCh38
rs778558550 Chr.3: 128910763-128910763 on Build GRCh38
rs781042917 Chr.3: 128910732-128910732 on Build GRCh38
rs186196906 Chr.3: 128910866-128910866 on Build GRCh38
rs749152088 Chr.3: 128910758-128910758 on Build GRCh38
rs764208038 Chr.3: 128910826-128910826 on Build GRCh38
rs201466811 Chr.3: 128910844-128910844 on Build GRCh38
rs200304794 Chr.3: 128910831-128910831 on Build GRCh38
rs138264216 Chr.3: 128910765-128910765 on Build GRCh38
rs777218042 Chr.3: 128910801-128910801 on Build GRCh38
rs769529395 Chr.3: 128910753-128910753 on Build GRCh38
rs34085303 Chr.3: 128910840-128910841 on Build GRCh38
rs758845674 Chr.3: 128910823-128910823 on Build GRCh38
rs758066233 Chr.3: 128910809-128910809 on Build GRCh38
rs768560744 Chr.3: 128910859-128910859 on Build GRCh38
rs749025449 Chr.3: 128910850-128910850 on Build GRCh38
rs752972035 Chr.3: 128910820-128910820 on Build GRCh38
rs779489971 Chr.3: 128910884-128910884 on Build GRCh38
rs369160539 Chr.3: 128910697-128910697 on Build GRCh38

View Full Product Details