Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 151363894-151364159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101929319
Entrez Gene ID
101929319
Gene Name
uncharacterized LOC101929319
Gene Aliases
-
UniGene
Hs.639241
Gene Chromosome Location
Chr.2: 151337826-151379371 on Build GRCh38
Transcripts
NR_110248.1 NM_007115.3

Gene Symbol
TNFAIP6
Entrez Gene ID
7130
Gene Name
TNF alpha induced protein 6
Gene Aliases
TSG-6, TSG6
UniGene
Hs.437322
Gene Chromosome Location
Chr.2: 151357592-151380048 on Build GRCh38
Transcripts
NR_110248.1 NM_007115.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4391123 c.208C>T p.Q70* Chr.2: 151364056-151364056 on Build GRCh38
6154239 c.167C>A p.A56E Chr.2: 151364015-151364015 on Build GRCh38
3425281 c.160G>A p.A54T Chr.2: 151364008-151364008 on Build GRCh38
6154238 c.208C>A p.Q70K Chr.2: 151364056-151364056 on Build GRCh38
475997 c.123A>C p.E41D Chr.2: 151363971-151363971 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557940940 Chr.2: 151364001-151364001 on Build GRCh38
rs577924895 Chr.2: 151364002-151364002 on Build GRCh38
rs780008246 Chr.2: 151364130-151364130 on Build GRCh38
rs375585000 Chr.2: 151363992-151363992 on Build GRCh38
rs868652351 Chr.2: 151364038-151364038 on Build GRCh38
rs751976860 Chr.2: 151363935-151363935 on Build GRCh38
rs767039176 Chr.2: 151364053-151364053 on Build GRCh38
rs756155042 Chr.2: 151364096-151364096 on Build GRCh38
rs758311675 Chr.2: 151364118-151364118 on Build GRCh38
rs760436617 Chr.2: 151364035-151364035 on Build GRCh38
rs758767494 Chr.2: 151363963-151363963 on Build GRCh38
rs752879888 Chr.2: 151364095-151364095 on Build GRCh38
rs1002258799 Chr.2: 151364065-151364065 on Build GRCh38
rs761269991 Chr.2: 151363946-151363946 on Build GRCh38
rs373889474 Chr.2: 151364056-151364056 on Build GRCh38
rs775252689 Chr.2: 151364034-151364034 on Build GRCh38
rs368107504 Chr.2: 151364105-151364105 on Build GRCh38
rs775060068 Chr.2: 151363926-151363926 on Build GRCh38
rs755232588 Chr.2: 151364043-151364043 on Build GRCh38
rs767536779 Chr.2: 151363928-151363928 on Build GRCh38
rs768323812 Chr.2: 151364044-151364044 on Build GRCh38
rs780559171 Chr.2: 151363974-151363974 on Build GRCh38
rs75961064 Chr.2: 151363996-151363996 on Build GRCh38
rs138376471 Chr.2: 151364041-151364041 on Build GRCh38
rs557244131 Chr.2: 151364114-151364114 on Build GRCh38
rs765725814 Chr.2: 151363953-151363953 on Build GRCh38
rs938301739 Chr.2: 151364083-151364083 on Build GRCh38
rs778767210 Chr.2: 151364012-151364012 on Build GRCh38
rs150253672 Chr.2: 151364050-151364050 on Build GRCh38
rs763712766 Chr.2: 151363945-151363945 on Build GRCh38
rs750979012 Chr.2: 151363954-151363954 on Build GRCh38
rs201398199 Chr.2: 151363978-151363978 on Build GRCh38
rs148912042 Chr.2: 151363982-151363982 on Build GRCh38
rs781430670 Chr.2: 151364094-151364094 on Build GRCh38
rs372855249 Chr.2: 151364014-151364014 on Build GRCh38
rs748269349 Chr.2: 151364126-151364126 on Build GRCh38
rs755326330 Chr.2: 151364064-151364064 on Build GRCh38
rs763538732 Chr.2: 151364052-151364052 on Build GRCh38
rs1046624496 Chr.2: 151364007-151364007 on Build GRCh38
rs369173073 Chr.2: 151364016-151364016 on Build GRCh38
rs750019197 Chr.2: 151364051-151364051 on Build GRCh38
rs566948911 Chr.2: 151363976-151363976 on Build GRCh38
rs776169770 Chr.2: 151364048-151364048 on Build GRCh38
rs781509964 Chr.2: 151364125-151364126 on Build GRCh38

View Full Product Details