Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99318688-99318951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH1B
Entrez Gene ID
125
Gene Name
alcohol dehydrogenase 1B (class I), beta polypeptide
Gene Aliases
ADH2, HEL-S-117
UniGene
Hs.4
Gene Chromosome Location
Chr.4: 99306387-99321442 on Build GRCh38
Transcripts
NM_000668.5 NM_001286650.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1618193 c.120+1G>A p.? Chr.4: 99318784-99318784 on Build GRCh38
3665424 c.107_120del14 p.E36fs*11 Chr.4: 99318785-99318798 on Build GRCh38
2951312 c.112C>T p.R38C Chr.4: 99318793-99318793 on Build GRCh38
3664638 c.85G>T p.V29F Chr.4: 99318820-99318820 on Build GRCh38
480670 c.114C>G p.R38R Chr.4: 99318791-99318791 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781090391 Chr.4: 99318739-99318739 on Build GRCh38
rs771467302 Chr.4: 99318896-99318896 on Build GRCh38
rs755834727 Chr.4: 99318744-99318744 on Build GRCh38
rs1037809204 Chr.4: 99318832-99318832 on Build GRCh38
rs779569999 Chr.4: 99318751-99318751 on Build GRCh38
rs770117532 Chr.4: 99318793-99318793 on Build GRCh38
rs775558705 Chr.4: 99318795-99318795 on Build GRCh38
rs761102982 Chr.4: 99318843-99318843 on Build GRCh38
rs758986990 Chr.4: 99318763-99318763 on Build GRCh38
rs778258146 Chr.4: 99318775-99318775 on Build GRCh38
rs767328444 Chr.4: 99318831-99318831 on Build GRCh38
rs906020550 Chr.4: 99318842-99318842 on Build GRCh38
rs1229983 Chr.4: 99318845-99318845 on Build GRCh38
rs200481568 Chr.4: 99318790-99318790 on Build GRCh38
rs771307803 Chr.4: 99318791-99318791 on Build GRCh38
rs777100313 Chr.4: 99318792-99318792 on Build GRCh38
rs78501598 Chr.4: 99318784-99318784 on Build GRCh38
rs746299652 Chr.4: 99318911-99318911 on Build GRCh38
rs374975751 Chr.4: 99318854-99318854 on Build GRCh38
rs778345918 Chr.4: 99318861-99318861 on Build GRCh38
rs753422844 Chr.4: 99318850-99318850 on Build GRCh38
rs201450842 Chr.4: 99318764-99318764 on Build GRCh38
rs368357625 Chr.4: 99318914-99318914 on Build GRCh38
rs555863882 Chr.4: 99318804-99318804 on Build GRCh38
rs201421877 Chr.4: 99318809-99318809 on Build GRCh38
rs765192867 Chr.4: 99318772-99318772 on Build GRCh38
rs113075608 Chr.4: 99318859-99318859 on Build GRCh38
rs72552716 Chr.4: 99318855-99318856 on Build GRCh38
rs55963727 Chr.4: 99318907-99318907 on Build GRCh38
rs375965352 Chr.4: 99318893-99318893 on Build GRCh38
rs750301947 Chr.4: 99318834-99318834 on Build GRCh38
rs763134176 Chr.4: 99318799-99318799 on Build GRCh38
rs775325816 Chr.4: 99318814-99318814 on Build GRCh38
rs773106812 Chr.4: 99318900-99318900 on Build GRCh38
rs747473547 Chr.4: 99318776-99318776 on Build GRCh38
rs28913913 Chr.4: 99318741-99318741 on Build GRCh38
rs751573568 Chr.4: 99318806-99318806 on Build GRCh38

View Full Product Details