Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 74888148-74888377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR59
Entrez Gene ID
79726
Gene Name
WD repeat domain 59
Gene Aliases
FP977
UniGene
Hs.280951
Gene Chromosome Location
Chr.16: 74873568-74985146 on Build GRCh38
Transcripts
NM_030581.3 XM_005256146.3 XM_011523331.2 XM_011523332.2 XM_017023667.1 XM_017023668.1 XM_017023669.1 XM_017023670.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5550470 c.2237C>T p.A746V Chr.16: 74888278-74888278 on Build GRCh38
3726271 c.2214G>T p.R738R Chr.16: 74888301-74888301 on Build GRCh38
6282512 c.2328G>T p.V776V Chr.16: 74888187-74888187 on Build GRCh38
5695561 c.2265_2288del24 p.Q755_N763>H Chr.16: 74888227-74888250 on Build GRCh38
7259654 c.2312G>A p.R771H Chr.16: 74888203-74888203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774264987 Chr.16: 74888189-74888189 on Build GRCh38
rs373676230 Chr.16: 74888246-74888246 on Build GRCh38
rs140276713 Chr.16: 74888278-74888278 on Build GRCh38
rs766972429 Chr.16: 74888301-74888301 on Build GRCh38
rs557062171 Chr.16: 74888283-74888283 on Build GRCh38
rs553271604 Chr.16: 74888309-74888309 on Build GRCh38
rs201398375 Chr.16: 74888284-74888284 on Build GRCh38
rs770808575 Chr.16: 74888178-74888178 on Build GRCh38
rs771051855 Chr.16: 74888272-74888272 on Build GRCh38
rs761933462 Chr.16: 74888203-74888203 on Build GRCh38
rs763073514 Chr.16: 74888180-74888180 on Build GRCh38
rs13339273 Chr.16: 74888333-74888333 on Build GRCh38
rs200182893 Chr.16: 74888177-74888177 on Build GRCh38
rs767610772 Chr.16: 74888204-74888204 on Build GRCh38
rs759506239 Chr.16: 74888264-74888264 on Build GRCh38
rs764444848 Chr.16: 74888181-74888181 on Build GRCh38
rs920632490 Chr.16: 74888353-74888353 on Build GRCh38
rs374585960 Chr.16: 74888342-74888342 on Build GRCh38
rs757111653 Chr.16: 74888330-74888330 on Build GRCh38
rs546843724 Chr.16: 74888225-74888225 on Build GRCh38
rs754577610 Chr.16: 74888237-74888237 on Build GRCh38
rs147955820 Chr.16: 74888320-74888320 on Build GRCh38
rs1050023152 Chr.16: 74888184-74888184 on Build GRCh38
rs138261959 Chr.16: 74888232-74888232 on Build GRCh38
rs931220165 Chr.16: 74888176-74888176 on Build GRCh38
rs769595260 Chr.16: 74888341-74888341 on Build GRCh38
rs762056077 Chr.16: 74888291-74888291 on Build GRCh38
rs755993642 Chr.16: 74888308-74888308 on Build GRCh38
rs758126318 Chr.16: 74888312-74888312 on Build GRCh38
rs746887756 Chr.16: 74888268-74888268 on Build GRCh38
rs9940422 Chr.16: 74888208-74888208 on Build GRCh38
rs552331119 Chr.16: 74888265-74888265 on Build GRCh38
rs147132881 Chr.16: 74888245-74888245 on Build GRCh38
rs199518523 Chr.16: 74888277-74888277 on Build GRCh38
rs144238663 Chr.16: 74888302-74888302 on Build GRCh38
rs748313800 Chr.16: 74888352-74888352 on Build GRCh38
rs1021256607 Chr.16: 74888269-74888269 on Build GRCh38
rs755419556 Chr.16: 74888303-74888303 on Build GRCh38
rs530156818 Chr.16: 74888217-74888217 on Build GRCh38
rs777410895 Chr.16: 74888319-74888319 on Build GRCh38
rs767371477 Chr.16: 74888296-74888296 on Build GRCh38
rs143856438 Chr.16: 74888271-74888271 on Build GRCh38
rs200598829 Chr.16: 74888250-74888250 on Build GRCh38
rs758187036 Chr.16: 74888319-74888319 on Build GRCh38
rs776286734 Chr.16: 74888275-74888275 on Build GRCh38
rs962128772 Chr.16: 74888332-74888332 on Build GRCh38
rs761154451 Chr.16: 74888297-74888297 on Build GRCh38

View Full Product Details