Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128805940-128806177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D13
Entrez Gene ID
54662
Gene Name
TBC1 domain family member 13
Gene Aliases
-
UniGene
Hs.3376
Gene Chromosome Location
Chr.9: 128787207-128810432 on Build GRCh38
Transcripts
NM_001286772.1 NM_018201.4 XM_005252060.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5584144 c.1005C>T p.I335I Chr.9: 128805945-128805945 on Build GRCh38
203914 c.1068C>T p.C356C Chr.9: 128806008-128806008 on Build GRCh38
6550508 c.1000G>T p.V334F Chr.9: 128805940-128805940 on Build GRCh38
7275905 c.1029C>T p.A343A Chr.9: 128805969-128805969 on Build GRCh38
3904581 c.1037A>G p.N346S Chr.9: 128805977-128805977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756945992 Chr.9: 128805993-128805993 on Build GRCh38
rs751519216 Chr.9: 128805987-128805987 on Build GRCh38
rs750224476 Chr.9: 128806000-128806000 on Build GRCh38
rs1054837909 Chr.9: 128805986-128805986 on Build GRCh38
rs758042718 Chr.9: 128805972-128805972 on Build GRCh38
rs759266956 Chr.9: 128805968-128805968 on Build GRCh38
rs376578779 Chr.9: 128805999-128805999 on Build GRCh38
rs556676692 Chr.9: 128806053-128806053 on Build GRCh38
rs764123425 Chr.9: 128805980-128805980 on Build GRCh38
rs2417127 Chr.9: 128806084-128806084 on Build GRCh38
rs769561824 Chr.9: 128806049-128806049 on Build GRCh38
rs147552518 Chr.9: 128806093-128806093 on Build GRCh38
rs963251068 Chr.9: 128806127-128806127 on Build GRCh38
rs763711785 Chr.9: 128806064-128806064 on Build GRCh38
rs772281535 Chr.9: 128806030-128806030 on Build GRCh38
rs780313149 Chr.9: 128806005-128806005 on Build GRCh38
rs558699933 Chr.9: 128806095-128806095 on Build GRCh38
rs1016946366 Chr.9: 128806119-128806119 on Build GRCh38
rs376635180 Chr.9: 128805966-128805966 on Build GRCh38
rs985420835 Chr.9: 128806116-128806116 on Build GRCh38
rs765024773 Chr.9: 128805969-128805969 on Build GRCh38
rs541006590 Chr.9: 128806156-128806156 on Build GRCh38
rs372896776 Chr.9: 128806027-128806027 on Build GRCh38
rs1051594507 Chr.9: 128806070-128806070 on Build GRCh38
rs768723863 Chr.9: 128806018-128806018 on Build GRCh38
rs73669976 Chr.9: 128806052-128806052 on Build GRCh38
rs572206576 Chr.9: 128806096-128806096 on Build GRCh38
rs199968908 Chr.9: 128805970-128805970 on Build GRCh38
rs1012258178 Chr.9: 128806147-128806147 on Build GRCh38
rs940732371 Chr.9: 128806090-128806090 on Build GRCh38
rs749364781 Chr.9: 128806009-128806009 on Build GRCh38
rs867049466 Chr.9: 128805991-128805991 on Build GRCh38
rs374312643 Chr.9: 128806039-128806039 on Build GRCh38
rs370685443 Chr.9: 128806028-128806028 on Build GRCh38
rs773597555 Chr.9: 128806036-128806036 on Build GRCh38
rs1037802932 Chr.9: 128806091-128806091 on Build GRCh38
rs756838115 Chr.9: 128805989-128805991 on Build GRCh38

View Full Product Details