Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 48221212-48221473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA13
Entrez Gene ID
154664
Gene Name
ATP binding cassette subfamily A member 13
Gene Aliases
-
UniGene
Hs.226568
Gene Chromosome Location
Chr.7: 48171424-48647497 on Build GRCh38
Transcripts
NM_152701.4 XM_011515130.2 XM_011515131.2 XM_011515132.2 XM_011515133.2 XM_011515134.2 XM_011515137.2 XM_011515138.2 XM_011515139.2 XM_011515141.2 XM_011515142.2 XM_011515143.2 XM_011515144.2 XM_011515145.2 XM_011515146.2 XM_011515147.2 XM_011515148.2 XM_011515149.2 XM_017011767.1 XM_017011768.1 XM_017011769.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1451156 c.293_294insT p.T101fs*6 Chr.7: 48221299-48221300 on Build GRCh38
3264124 c.459delT p.F155fs*11 Chr.7: 48221300-48221300 on Build GRCh38
5573236 c.275-1G>A p.? Chr.7: 48221280-48221280 on Build GRCh38
5438021 c.467C>A p.T156K Chr.7: 48221308-48221308 on Build GRCh38
1451155 c.458_459insT p.T156fs*6 Chr.7: 48221299-48221300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531073745 Chr.7: 48221326-48221326 on Build GRCh38
rs774227804 Chr.7: 48221354-48221354 on Build GRCh38
rs370823905 Chr.7: 48221297-48221297 on Build GRCh38
rs533616376 Chr.7: 48221439-48221439 on Build GRCh38
rs937419124 Chr.7: 48221324-48221324 on Build GRCh38
rs997992355 Chr.7: 48221425-48221425 on Build GRCh38
rs1011192091 Chr.7: 48221418-48221418 on Build GRCh38
rs905837440 Chr.7: 48221322-48221322 on Build GRCh38
rs930836287 Chr.7: 48221292-48221292 on Build GRCh38
rs908153160 Chr.7: 48221299-48221299 on Build GRCh38
rs1016221855 Chr.7: 48221449-48221449 on Build GRCh38
rs36072375 Chr.7: 48221428-48221428 on Build GRCh38
rs950295186 Chr.7: 48221312-48221312 on Build GRCh38
rs762976281 Chr.7: 48221254-48221254 on Build GRCh38
rs747685045 Chr.7: 48221239-48221239 on Build GRCh38
rs10252745 Chr.7: 48221272-48221272 on Build GRCh38
rs751115699 Chr.7: 48221371-48221371 on Build GRCh38
rs900689624 Chr.7: 48221423-48221423 on Build GRCh38
rs116812541 Chr.7: 48221335-48221335 on Build GRCh38
rs1046146921 Chr.7: 48221315-48221315 on Build GRCh38
rs886432042 Chr.7: 48221429-48221429 on Build GRCh38
rs200795824 Chr.7: 48221250-48221250 on Build GRCh38

View Full Product Details