Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 192658160-192658405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RGS13
Entrez Gene ID
6003
Gene Name
regulator of G-protein signaling 13
Gene Aliases
-
UniGene
Hs.497220
Gene Chromosome Location
Chr.1: 192636138-192660311 on Build GRCh38
Transcripts
NM_002927.4 NM_144766.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5802922 c.227G>A p.R76Q Chr.1: 192658300-192658300 on Build GRCh38
5803476 c.135A>T p.P45P Chr.1: 192658208-192658208 on Build GRCh38
901346 c.252G>T p.K84N Chr.1: 192658325-192658325 on Build GRCh38
901347 c.268T>G p.Y90D Chr.1: 192658341-192658341 on Build GRCh38
5723511 c.267T>A p.I89I Chr.1: 192658340-192658340 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779965280 Chr.1: 192658239-192658239 on Build GRCh38
rs372688055 Chr.1: 192658223-192658223 on Build GRCh38
rs968118519 Chr.1: 192658336-192658336 on Build GRCh38
rs747673405 Chr.1: 192658344-192658344 on Build GRCh38
rs747390065 Chr.1: 192658366-192658366 on Build GRCh38
rs761040791 Chr.1: 192658193-192658193 on Build GRCh38
rs758497365 Chr.1: 192658292-192658292 on Build GRCh38
rs757076213 Chr.1: 192658213-192658213 on Build GRCh38
rs767057646 Chr.1: 192658265-192658265 on Build GRCh38
rs527971820 Chr.1: 192658358-192658358 on Build GRCh38
rs143481310 Chr.1: 192658207-192658207 on Build GRCh38
rs761071258 Chr.1: 192658263-192658263 on Build GRCh38
rs780805183 Chr.1: 192658218-192658218 on Build GRCh38
rs199577739 Chr.1: 192658320-192658320 on Build GRCh38
rs750255257 Chr.1: 192658295-192658295 on Build GRCh38
rs1002501224 Chr.1: 192658234-192658234 on Build GRCh38
rs761333079 Chr.1: 192658256-192658256 on Build GRCh38
rs41300867 Chr.1: 192658301-192658301 on Build GRCh38
rs746628415 Chr.1: 192658242-192658242 on Build GRCh38
rs149567175 Chr.1: 192658208-192658208 on Build GRCh38
rs766971831 Chr.1: 192658200-192658200 on Build GRCh38
rs765337505 Chr.1: 192658280-192658280 on Build GRCh38
rs568720010 Chr.1: 192658317-192658317 on Build GRCh38
rs139968001 Chr.1: 192658240-192658240 on Build GRCh38
rs12401867 Chr.1: 192658288-192658288 on Build GRCh38
rs28531705 Chr.1: 192658267-192658267 on Build GRCh38
rs771362925 Chr.1: 192658375-192658375 on Build GRCh38
rs778346180 Chr.1: 192658327-192658327 on Build GRCh38
rs768100083 Chr.1: 192658247-192658247 on Build GRCh38
rs117958126 Chr.1: 192658268-192658268 on Build GRCh38
rs759701579 Chr.1: 192658279-192658279 on Build GRCh38
rs774000220 Chr.1: 192658248-192658248 on Build GRCh38
rs779898260 Chr.1: 192658226-192658226 on Build GRCh38
rs186216817 Chr.1: 192658357-192658357 on Build GRCh38
rs529030881 Chr.1: 192658299-192658299 on Build GRCh38
rs771664361 Chr.1: 192658354-192658354 on Build GRCh38
rs541050160 Chr.1: 192658259-192658259 on Build GRCh38
rs377283074 Chr.1: 192658318-192658318 on Build GRCh38
rs748857400 Chr.1: 192658246-192658246 on Build GRCh38
rs367819787 Chr.1: 192658211-192658211 on Build GRCh38
rs769665832 Chr.1: 192658233-192658233 on Build GRCh38
rs772510233 Chr.1: 192658258-192658258 on Build GRCh38
rs754009730 Chr.1: 192658202-192658202 on Build GRCh38
rs752908188 Chr.1: 192658210-192658210 on Build GRCh38

View Full Product Details