Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 17616687-17616938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRBP1
Entrez Gene ID
6238
Gene Name
ribosome binding protein 1
Gene Aliases
ES/130, ES130, RRp, hES
UniGene
Hs.472213
Gene Chromosome Location
Chr.20: 17613678-17682283 on Build GRCh38
Transcripts
NM_001042576.1 NM_004587.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4001838 c.2461-7G>A p.? Chr.20: 17616846-17616846 on Build GRCh38
1410546 c.3861C>T p.P1287P Chr.20: 17616738-17616738 on Build GRCh38
6847540 c.2507A>T p.D836V Chr.20: 17616793-17616793 on Build GRCh38
5904660 c.2524G>A p.A842T Chr.20: 17616776-17616776 on Build GRCh38
1024806 c.3840G>A p.A1280A Chr.20: 17616759-17616759 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs56102376 Chr.20: 17616724-17616724 on Build GRCh38
rs375371043 Chr.20: 17616918-17616918 on Build GRCh38
rs761553694 Chr.20: 17616917-17616917 on Build GRCh38
rs748600528 Chr.20: 17616814-17616814 on Build GRCh38
rs8183329 Chr.20: 17616846-17616846 on Build GRCh38
rs372942674 Chr.20: 17616887-17616887 on Build GRCh38
rs375442920 Chr.20: 17616729-17616729 on Build GRCh38
rs559756601 Chr.20: 17616810-17616810 on Build GRCh38
rs776275268 Chr.20: 17616760-17616760 on Build GRCh38
rs769876570 Chr.20: 17616723-17616723 on Build GRCh38
rs769272121 Chr.20: 17616738-17616738 on Build GRCh38
rs756414565 Chr.20: 17616796-17616796 on Build GRCh38
rs549359383 Chr.20: 17616915-17616915 on Build GRCh38
rs557939239 Chr.20: 17616763-17616763 on Build GRCh38
rs377270303 Chr.20: 17616881-17616881 on Build GRCh38
rs371810910 Chr.20: 17616712-17616712 on Build GRCh38
rs764003798 Chr.20: 17616852-17616852 on Build GRCh38
rs765211498 Chr.20: 17616772-17616772 on Build GRCh38
rs774967763 Chr.20: 17616747-17616747 on Build GRCh38
rs759112999 Chr.20: 17616842-17616842 on Build GRCh38
rs1056356239 Chr.20: 17616908-17616908 on Build GRCh38
rs878862183 Chr.20: 17616841-17616841 on Build GRCh38
rs201211533 Chr.20: 17616758-17616758 on Build GRCh38
rs754991097 Chr.20: 17616807-17616807 on Build GRCh38
rs752533144 Chr.20: 17616722-17616722 on Build GRCh38
rs756151022 Chr.20: 17616731-17616731 on Build GRCh38
rs11551705 Chr.20: 17616801-17616801 on Build GRCh38
rs141635452 Chr.20: 17616805-17616805 on Build GRCh38
rs41290441 Chr.20: 17616837-17616837 on Build GRCh38
rs566098133 Chr.20: 17616757-17616757 on Build GRCh38
rs772579605 Chr.20: 17616815-17616815 on Build GRCh38
rs976303905 Chr.20: 17616789-17616789 on Build GRCh38
rs200224134 Chr.20: 17616749-17616749 on Build GRCh38
rs545898145 Chr.20: 17616848-17616848 on Build GRCh38
rs147621443 Chr.20: 17616840-17616840 on Build GRCh38
rs576543669 Chr.20: 17616791-17616791 on Build GRCh38
rs752578231 Chr.20: 17616781-17616781 on Build GRCh38
rs547251626 Chr.20: 17616806-17616806 on Build GRCh38
rs763808399 Chr.20: 17616782-17616782 on Build GRCh38
rs34479552 Chr.20: 17616737-17616737 on Build GRCh38
rs755004351 Chr.20: 17616715-17616715 on Build GRCh38
rs554331772 Chr.20: 17616752-17616752 on Build GRCh38
rs751437079 Chr.20: 17616871-17616871 on Build GRCh38
rs375724670 Chr.20: 17616826-17616826 on Build GRCh38
rs147915375 Chr.20: 17616750-17616750 on Build GRCh38
rs139695441 Chr.20: 17616733-17616733 on Build GRCh38
rs911348 Chr.20: 17616716-17616716 on Build GRCh38
rs778365203 Chr.20: 17616878-17616879 on Build GRCh38
rs2236252 Chr.20: 17616886-17616886 on Build GRCh38
rs371306215 Chr.20: 17616759-17616759 on Build GRCh38
rs572121494 Chr.20: 17616847-17616847 on Build GRCh38
rs531233694 Chr.20: 17616890-17616890 on Build GRCh38

View Full Product Details