Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 44949947-44950204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SETBP1
Entrez Gene ID
26040
Gene Name
SET binding protein 1
Gene Aliases
MRD29, SEB
UniGene
Hs.435458
Gene Chromosome Location
Chr.18: 44680173-45068510 on Build GRCh38
Transcripts
NM_015559.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6524191 c.807T>A p.S269R Chr.18: 44950147-44950147 on Build GRCh38
3525849 c.799A>G p.K267E Chr.18: 44950139-44950139 on Build GRCh38
6498475 c.812G>C p.G271A Chr.18: 44950152-44950152 on Build GRCh38
6524190 c.789C>A p.A263A Chr.18: 44950129-44950129 on Build GRCh38
1729986 c.779T>C p.F260S Chr.18: 44950119-44950119 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776263750 Chr.18: 44950038-44950038 on Build GRCh38
rs774801388 Chr.18: 44950093-44950093 on Build GRCh38
rs749637523 Chr.18: 44950059-44950059 on Build GRCh38
rs758270974 Chr.18: 44950132-44950132 on Build GRCh38
rs753661124 Chr.18: 44950017-44950017 on Build GRCh38
rs764663303 Chr.18: 44949996-44949996 on Build GRCh38
rs994741648 Chr.18: 44950120-44950120 on Build GRCh38
rs768953472 Chr.18: 44950153-44950153 on Build GRCh38
rs1002734293 Chr.18: 44950171-44950171 on Build GRCh38
rs556231993 Chr.18: 44949995-44949995 on Build GRCh38
rs749986583 Chr.18: 44950010-44950010 on Build GRCh38
rs770504929 Chr.18: 44950035-44950035 on Build GRCh38
rs755762478 Chr.18: 44950011-44950011 on Build GRCh38
rs35468095 Chr.18: 44950153-44950154 on Build GRCh38
rs766190985 Chr.18: 44950015-44950015 on Build GRCh38
rs1009862226 Chr.18: 44949976-44949976 on Build GRCh38
rs766021423 Chr.18: 44950105-44950105 on Build GRCh38
rs747982980 Chr.18: 44950028-44950028 on Build GRCh38
rs764797231 Chr.18: 44950182-44950182 on Build GRCh38
rs760327515 Chr.18: 44950101-44950101 on Build GRCh38
rs756231270 Chr.18: 44950142-44950142 on Build GRCh38
rs761385178 Chr.18: 44950005-44950005 on Build GRCh38
rs377615601 Chr.18: 44950030-44950030 on Build GRCh38
rs1050167249 Chr.18: 44950076-44950076 on Build GRCh38
rs745946867 Chr.18: 44950141-44950141 on Build GRCh38
rs777748383 Chr.18: 44950134-44950134 on Build GRCh38
rs149073567 Chr.18: 44950024-44950024 on Build GRCh38
rs749422908 Chr.18: 44950152-44950152 on Build GRCh38
rs752490647 Chr.18: 44950128-44950128 on Build GRCh38
rs773892934 Chr.18: 44950177-44950177 on Build GRCh38
rs886053788 Chr.18: 44950156-44950156 on Build GRCh38
rs143196787 Chr.18: 44950025-44950025 on Build GRCh38
rs972701994 Chr.18: 44950131-44950131 on Build GRCh38
rs774764842 Chr.18: 44950000-44950000 on Build GRCh38
rs760239764 Chr.18: 44950179-44950179 on Build GRCh38
rs149472556 Chr.18: 44950097-44950097 on Build GRCh38
rs11082414 Chr.18: 44950031-44950031 on Build GRCh38
rs770005446 Chr.18: 44949977-44949977 on Build GRCh38
rs767087990 Chr.18: 44950009-44950009 on Build GRCh38
rs79615803 Chr.18: 44949974-44949974 on Build GRCh38
rs768953971 Chr.18: 44950083-44950083 on Build GRCh38
rs770296814 Chr.18: 44950183-44950183 on Build GRCh38
rs780304631 Chr.18: 44950148-44950148 on Build GRCh38
rs772700161 Chr.18: 44950159-44950159 on Build GRCh38
rs1027926542 Chr.18: 44950146-44950146 on Build GRCh38
rs769993210 Chr.18: 44950044-44950044 on Build GRCh38
rs141858699 Chr.18: 44950108-44950108 on Build GRCh38
rs748537549 Chr.18: 44950158-44950158 on Build GRCh38
rs202152985 Chr.18: 44949985-44949985 on Build GRCh38
rs775644229 Chr.18: 44950053-44950053 on Build GRCh38
rs764896608 Chr.18: 44950121-44950121 on Build GRCh38
rs144967125 Chr.18: 44950109-44950109 on Build GRCh38
rs774513800 Chr.18: 44950157-44950157 on Build GRCh38

View Full Product Details