Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 131163033-131163302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCERG1L
Entrez Gene ID
256536
Gene Name
transcription elongation regulator 1 like
Gene Aliases
-
UniGene
Hs.126575
Gene Chromosome Location
Chr.10: 131092391-131311721 on Build GRCh38
Transcripts
NM_174937.3 XM_011539577.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4815735 c.1016C>G p.P339R Chr.10: 131163140-131163140 on Build GRCh38
4845476 c.848G>T p.G283V Chr.10: 131163185-131163185 on Build GRCh38
4456970 c.906C>A p.S302S Chr.10: 131163127-131163127 on Build GRCh38
4845475 c.971G>T p.G324V Chr.10: 131163185-131163185 on Build GRCh38
401133 c.906C>G p.S302S Chr.10: 131163127-131163127 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763275423 Chr.10: 131163235-131163235 on Build GRCh38
rs759673435 Chr.10: 131163214-131163214 on Build GRCh38
rs768659906 Chr.10: 131163103-131163103 on Build GRCh38
rs764547795 Chr.10: 131163180-131163180 on Build GRCh38
rs753001245 Chr.10: 131163163-131163163 on Build GRCh38
rs776280302 Chr.10: 131163223-131163224 on Build GRCh38
rs17857275 Chr.10: 131163140-131163140 on Build GRCh38
rs913358266 Chr.10: 131163110-131163110 on Build GRCh38
rs201354515 Chr.10: 131163200-131163200 on Build GRCh38
rs1008783142 Chr.10: 131163220-131163220 on Build GRCh38
rs764491048 Chr.10: 131163236-131163236 on Build GRCh38
rs183908854 Chr.10: 131163208-131163208 on Build GRCh38
rs372921989 Chr.10: 131163085-131163085 on Build GRCh38
rs118074196 Chr.10: 131163191-131163191 on Build GRCh38
rs1024032423 Chr.10: 131163231-131163231 on Build GRCh38
rs765326383 Chr.10: 131163232-131163232 on Build GRCh38
rs575264687 Chr.10: 131163060-131163060 on Build GRCh38
rs756770983 Chr.10: 131163183-131163183 on Build GRCh38
rs753602439 Chr.10: 131163133-131163133 on Build GRCh38
rs766567251 Chr.10: 131163258-131163258 on Build GRCh38
rs868451954 Chr.10: 131163125-131163125 on Build GRCh38
rs143934935 Chr.10: 131163193-131163193 on Build GRCh38
rs775218857 Chr.10: 131163088-131163088 on Build GRCh38
rs1042274599 Chr.10: 131163173-131163173 on Build GRCh38
rs148112736 Chr.10: 131163192-131163192 on Build GRCh38
rs993856737 Chr.10: 131163230-131163230 on Build GRCh38
rs776898038 Chr.10: 131163136-131163136 on Build GRCh38
rs780629059 Chr.10: 131163188-131163188 on Build GRCh38
rs765498738 Chr.10: 131163145-131163145 on Build GRCh38
rs748091063 Chr.10: 131163199-131163199 on Build GRCh38
rs368565982 Chr.10: 131163083-131163083 on Build GRCh38
rs774403927 Chr.10: 131163195-131163195 on Build GRCh38
rs1005479301 Chr.10: 131163094-131163094 on Build GRCh38
rs548626336 Chr.10: 131163170-131163170 on Build GRCh38
rs550346101 Chr.10: 131163181-131163181 on Build GRCh38
rs140546881 Chr.10: 131163260-131163260 on Build GRCh38
rs774458961 Chr.10: 131163121-131163121 on Build GRCh38
rs193104282 Chr.10: 131163139-131163139 on Build GRCh38
rs530447519 Chr.10: 131163084-131163084 on Build GRCh38
rs375514917 Chr.10: 131163102-131163102 on Build GRCh38
rs199863432 Chr.10: 131163233-131163233 on Build GRCh38

View Full Product Details