Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67631694-67631919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX10
Entrez Gene ID
347853
Gene Name
T-box 10
Gene Aliases
TBX13, TBX7
UniGene
Hs.454480
Gene Chromosome Location
Chr.11: 67631303-67639560 on Build GRCh38
Transcripts
NM_005995.4 XM_011544992.2 XM_011544993.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3980619 c.982C>G p.Q328E Chr.11: 67631781-67631781 on Build GRCh38
6320149 c.977C>A p.T326K Chr.11: 67631786-67631786 on Build GRCh38
6347328 c.1044C>A p.Y348* Chr.11: 67631719-67631719 on Build GRCh38
4036046 c.1005G>A p.P335P Chr.11: 67631758-67631758 on Build GRCh38
7321285 c.957G>A p.P319P Chr.11: 67631806-67631806 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761628835 Chr.11: 67631808-67631808 on Build GRCh38
rs775191487 Chr.11: 67631791-67631791 on Build GRCh38
rs758596700 Chr.11: 67631831-67631831 on Build GRCh38
rs370073114 Chr.11: 67631719-67631719 on Build GRCh38
rs915125877 Chr.11: 67631734-67631734 on Build GRCh38
rs531906539 Chr.11: 67631786-67631786 on Build GRCh38
rs767377721 Chr.11: 67631815-67631815 on Build GRCh38
rs755019737 Chr.11: 67631760-67631760 on Build GRCh38
rs748377405 Chr.11: 67631767-67631767 on Build GRCh38
rs201107050 Chr.11: 67631806-67631806 on Build GRCh38
rs374679098 Chr.11: 67631849-67631849 on Build GRCh38
rs1037963953 Chr.11: 67631891-67631891 on Build GRCh38
rs764900930 Chr.11: 67631716-67631716 on Build GRCh38
rs774916092 Chr.11: 67631896-67631896 on Build GRCh38
rs753876539 Chr.11: 67631759-67631759 on Build GRCh38
rs757697215 Chr.11: 67631863-67631863 on Build GRCh38
rs369161057 Chr.11: 67631825-67631825 on Build GRCh38
rs778321213 Chr.11: 67631773-67631773 on Build GRCh38
rs186569241 Chr.11: 67631876-67631876 on Build GRCh38
rs769321117 Chr.11: 67631888-67631888 on Build GRCh38
rs765281303 Chr.11: 67631869-67631869 on Build GRCh38
rs781132994 Chr.11: 67631755-67631755 on Build GRCh38
rs899406445 Chr.11: 67631895-67631895 on Build GRCh38
rs868037507 Chr.11: 67631861-67631861 on Build GRCh38
rs762746844 Chr.11: 67631733-67631733 on Build GRCh38
rs750374429 Chr.11: 67631818-67631818 on Build GRCh38
rs372881982 Chr.11: 67631828-67631828 on Build GRCh38
rs763930638 Chr.11: 67631797-67631797 on Build GRCh38
rs746415369 Chr.11: 67631887-67631887 on Build GRCh38
rs771514015 Chr.11: 67631779-67631779 on Build GRCh38
rs201389909 Chr.11: 67631729-67631729 on Build GRCh38
rs757238456 Chr.11: 67631749-67631749 on Build GRCh38
rs763866445 Chr.11: 67631737-67631737 on Build GRCh38
rs747335405 Chr.11: 67631860-67631860 on Build GRCh38
rs912442180 Chr.11: 67631756-67631756 on Build GRCh38
rs747494980 Chr.11: 67631777-67631777 on Build GRCh38
rs142856471 Chr.11: 67631785-67631785 on Build GRCh38
rs755072160 Chr.11: 67631820-67631820 on Build GRCh38
rs867198377 Chr.11: 67631893-67631893 on Build GRCh38
rs762557472 Chr.11: 67631795-67631795 on Build GRCh38
rs751471550 Chr.11: 67631738-67631738 on Build GRCh38
rs779139266 Chr.11: 67631761-67631761 on Build GRCh38
rs772479926 Chr.11: 67631771-67631771 on Build GRCh38

View Full Product Details