Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 42459706-42459965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPCS
Entrez Gene ID
79717
Gene Name
phosphopantothenoylcysteine synthetase
Gene Aliases
-
UniGene
Hs.706662
Gene Chromosome Location
Chr.1: 42456341-42473381 on Build GRCh38
Transcripts
NM_001077447.2 NM_001287506.1 NM_001287507.1 NM_001287508.1 NM_001287509.1 NM_001287510.1 NM_001287511.1 NM_024664.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7155399 c.860A>T p.K287I Chr.1: 42459850-42459850 on Build GRCh38
5567928 c.822G>A p.S274S Chr.1: 42459812-42459812 on Build GRCh38
3738232 c.741G>T p.L247F Chr.1: 42459731-42459731 on Build GRCh38
1639888 c.767T>C p.V256A Chr.1: 42459757-42459757 on Build GRCh38
3738082 c.742G>A p.E248K Chr.1: 42459732-42459732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753360321 Chr.1: 42459925-42459925 on Build GRCh38
rs961368391 Chr.1: 42459849-42459849 on Build GRCh38
rs752560159 Chr.1: 42459794-42459794 on Build GRCh38
rs375090300 Chr.1: 42459777-42459777 on Build GRCh38
rs541572840 Chr.1: 42459769-42459769 on Build GRCh38
rs763459889 Chr.1: 42459803-42459803 on Build GRCh38
rs781766081 Chr.1: 42459839-42459839 on Build GRCh38
rs755679446 Chr.1: 42459798-42459798 on Build GRCh38
rs779561295 Chr.1: 42459936-42459936 on Build GRCh38
rs780467070 Chr.1: 42459844-42459844 on Build GRCh38
rs916564907 Chr.1: 42459786-42459786 on Build GRCh38
rs1049600110 Chr.1: 42459812-42459812 on Build GRCh38
rs967199663 Chr.1: 42459751-42459751 on Build GRCh38
rs775424687 Chr.1: 42459906-42459906 on Build GRCh38
rs756903119 Chr.1: 42459811-42459811 on Build GRCh38
rs1021499805 Chr.1: 42459742-42459742 on Build GRCh38
rs61745591 Chr.1: 42459782-42459782 on Build GRCh38
rs182769462 Chr.1: 42459855-42459855 on Build GRCh38
rs751422458 Chr.1: 42459780-42459780 on Build GRCh38
rs753362802 Chr.1: 42459809-42459809 on Build GRCh38
rs146107417 Chr.1: 42459752-42459752 on Build GRCh38
rs774498065 Chr.1: 42459874-42459874 on Build GRCh38
rs767455402 Chr.1: 42459785-42459785 on Build GRCh38
rs755339358 Chr.1: 42459832-42459832 on Build GRCh38
rs142078638 Chr.1: 42459830-42459830 on Build GRCh38
rs760575381 Chr.1: 42459917-42459917 on Build GRCh38
rs769698249 Chr.1: 42459758-42459758 on Build GRCh38
rs763038291 Chr.1: 42459767-42459767 on Build GRCh38
rs756781299 Chr.1: 42459937-42459937 on Build GRCh38
rs368483468 Chr.1: 42459919-42459919 on Build GRCh38
rs759327365 Chr.1: 42459894-42459894 on Build GRCh38
rs146331223 Chr.1: 42459854-42459854 on Build GRCh38
rs952234267 Chr.1: 42459924-42459924 on Build GRCh38
rs749521343 Chr.1: 42459866-42459866 on Build GRCh38
rs747351660 Chr.1: 42459748-42459748 on Build GRCh38
rs773515366 Chr.1: 42459766-42459766 on Build GRCh38
rs945281570 Chr.1: 42459743-42459743 on Build GRCh38
rs889514468 Chr.1: 42459828-42459828 on Build GRCh38
rs748563516 Chr.1: 42459842-42459842 on Build GRCh38
rs761999964 Chr.1: 42459756-42459756 on Build GRCh38
rs767257417 Chr.1: 42459895-42459895 on Build GRCh38
rs1047645204 Chr.1: 42459861-42459861 on Build GRCh38
rs368077055 Chr.1: 42459778-42459778 on Build GRCh38

View Full Product Details