Assay Design



Gene Transcripts

Gene Symbol
ALK
Entrez Gene ID
238
Gene Name
anaplastic lymphoma receptor tyrosine kinase
Gene Aliases
CD246, NBLST3
Chromosome Location
Chr.2: 29192774 - 29921611 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004304.4 NP_004295.2 5 - 6 2238 68
GenBank mRNA AB209477.4 - 5 - 6 1775 68
EU788003.1 - 5 - 6 2193 68
EU788004.1 - 5 - 6 2193 68
EU788005.1 - 5 - 6 2193 68
EU788006.1 - 5 - 6 2193 68
EU788007.1 - 5 - 6 2193 68
EU788008.1 - 5 - 6 2193 68
EU788009.1 - 5 - 6 2193 68
EU788010.1 - 5 - 6 2193 68
FJ838786.1 - 5 - 6 2193 68
FJ838787.1 - 5 - 6 2193 68
GU128152.1 - 5 - 6 1286 68
GU128153.1 - 5 - 6 1286 68
GU128154.1 - 5 - 6 1286 68
GU128155.1 - 5 - 6 1286 68
GU128156.1 - 5 - 6 1286 68
GU128157.1 - 5 - 6 1286 68
U62540.1 - 5 - 6 2197 68
U66559.1 - 5 - 6 1434 68

View Full Product Details