Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 23084063-23084316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD93
Entrez Gene ID
22918
Gene Name
CD93 molecule
Gene Aliases
C1QR1, C1qR(P), C1qRP, CDw93, ECSM3, MXRA4, dJ737E23.1
UniGene
Hs.97199
Gene Chromosome Location
Chr.20: 23076554-23086340 on Build GRCh38
Transcripts
NM_012072.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4001854 c.1918A>G p.M640V Chr.20: 23084275-23084275 on Build GRCh38
1166243 c.1908G>T p.E636D Chr.20: 23084285-23084285 on Build GRCh38
5578263 c.1925A>G p.N642S Chr.20: 23084268-23084268 on Build GRCh38
4892663 c.1895C>T p.P632L Chr.20: 23084298-23084298 on Build GRCh38
1247735 c.1910G>A p.S637N Chr.20: 23084283-23084283 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144986131 Chr.20: 23084271-23084271 on Build GRCh38
rs747053249 Chr.20: 23084261-23084261 on Build GRCh38
rs41480746 Chr.20: 23084294-23084294 on Build GRCh38
rs375179604 Chr.20: 23084210-23084210 on Build GRCh38
rs41412346 Chr.20: 23084130-23084130 on Build GRCh38
rs540441474 Chr.20: 23084277-23084277 on Build GRCh38
rs946143573 Chr.20: 23084203-23084203 on Build GRCh38
rs748574833 Chr.20: 23084280-23084280 on Build GRCh38
rs748545154 Chr.20: 23084250-23084250 on Build GRCh38
rs536026143 Chr.20: 23084108-23084108 on Build GRCh38
rs759865390 Chr.20: 23084266-23084266 on Build GRCh38
rs567456931 Chr.20: 23084097-23084097 on Build GRCh38
rs571236134 Chr.20: 23084116-23084116 on Build GRCh38
rs11087413 Chr.20: 23084173-23084173 on Build GRCh38
rs766792396 Chr.20: 23084120-23084120 on Build GRCh38
rs968293276 Chr.20: 23084098-23084098 on Build GRCh38
rs758871913 Chr.20: 23084281-23084281 on Build GRCh38
rs546895677 Chr.20: 23084111-23084111 on Build GRCh38
rs377312437 Chr.20: 23084292-23084292 on Build GRCh38
rs978842967 Chr.20: 23084246-23084246 on Build GRCh38
rs372945964 Chr.20: 23084217-23084217 on Build GRCh38
rs764886512 Chr.20: 23084267-23084267 on Build GRCh38
rs201518186 Chr.20: 23084253-23084253 on Build GRCh38
rs763086077 Chr.20: 23084244-23084244 on Build GRCh38
rs976782233 Chr.20: 23084167-23084167 on Build GRCh38
rs554324618 Chr.20: 23084237-23084237 on Build GRCh38
rs775674185 Chr.20: 23084199-23084199 on Build GRCh38
rs193126584 Chr.20: 23084132-23084132 on Build GRCh38
rs41418351 Chr.20: 23084275-23084275 on Build GRCh38
rs900111509 Chr.20: 23084287-23084287 on Build GRCh38
rs755804228 Chr.20: 23084278-23084278 on Build GRCh38
rs200232408 Chr.20: 23084272-23084272 on Build GRCh38
rs762840267 Chr.20: 23084270-23084270 on Build GRCh38
rs772728442 Chr.20: 23084251-23084251 on Build GRCh38
rs955513861 Chr.20: 23084082-23084082 on Build GRCh38
rs752762254 Chr.20: 23084236-23084236 on Build GRCh38
rs776702004 Chr.20: 23084262-23084262 on Build GRCh38
rs575311304 Chr.20: 23084140-23084140 on Build GRCh38
rs1010080718 Chr.20: 23084091-23084091 on Build GRCh38
rs373581862 Chr.20: 23084109-23084109 on Build GRCh38
rs111435065 Chr.20: 23084124-23084124 on Build GRCh38
rs753827669 Chr.20: 23084293-23084293 on Build GRCh38
rs756254108 Chr.20: 23084249-23084249 on Build GRCh38

View Full Product Details