Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19635232-19635457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMIP
Entrez Gene ID
51291
Gene Name
GEM interacting protein
Gene Aliases
ARHGAP46
UniGene
Hs.49427
Gene Chromosome Location
Chr.19: 19629473-19643667 on Build GRCh38
Transcripts
NM_001288998.1 NM_001288999.1 NM_016573.3 XM_005259927.2 XM_011528048.2 XM_017026861.1 XM_017026862.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4000593 c.1560+10G>C p.? Chr.19: 19635405-19635405 on Build GRCh38
4075990 c.1524C>T p.C508C Chr.19: 19635451-19635451 on Build GRCh38
5789620 c.1548G>T p.T516T Chr.19: 19635427-19635427 on Build GRCh38
1744977 c.1543_1544GG>TT p.G515L Chr.19: 19635431-19635432 on Build GRCh38
1480826 c.1557G>A p.E519E Chr.19: 19635418-19635418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2304128 Chr.19: 19635342-19635342 on Build GRCh38
rs761162347 Chr.19: 19635433-19635433 on Build GRCh38
rs571239033 Chr.19: 19635302-19635302 on Build GRCh38
rs761227217 Chr.19: 19635427-19635427 on Build GRCh38
rs765086067 Chr.19: 19635413-19635413 on Build GRCh38
rs759340943 Chr.19: 19635378-19635378 on Build GRCh38
rs113619686 Chr.19: 19635320-19635320 on Build GRCh38
rs766881890 Chr.19: 19635434-19635434 on Build GRCh38
rs916669375 Chr.19: 19635289-19635289 on Build GRCh38
rs202225255 Chr.19: 19635432-19635432 on Build GRCh38
rs538009723 Chr.19: 19635271-19635271 on Build GRCh38
rs757221498 Chr.19: 19635388-19635388 on Build GRCh38
rs768140288 Chr.19: 19635414-19635414 on Build GRCh38
rs768702480 Chr.19: 19635397-19635397 on Build GRCh38
rs527487519 Chr.19: 19635260-19635260 on Build GRCh38
rs772696325 Chr.19: 19635428-19635428 on Build GRCh38
rs972479322 Chr.19: 19635353-19635353 on Build GRCh38
rs562554598 Chr.19: 19635271-19635272 on Build GRCh38
rs2304129 Chr.19: 19635405-19635405 on Build GRCh38
rs779072696 Chr.19: 19635265-19635265 on Build GRCh38
rs773835328 Chr.19: 19635430-19635430 on Build GRCh38
rs754083289 Chr.19: 19635254-19635254 on Build GRCh38
rs987038805 Chr.19: 19635328-19635328 on Build GRCh38
rs375796180 Chr.19: 19635371-19635371 on Build GRCh38
rs969479120 Chr.19: 19635300-19635300 on Build GRCh38
rs930258928 Chr.19: 19635391-19635391 on Build GRCh38
rs780984967 Chr.19: 19635392-19635392 on Build GRCh38
rs939859435 Chr.19: 19635351-19635351 on Build GRCh38
rs184488122 Chr.19: 19635291-19635291 on Build GRCh38
rs138031462 Chr.19: 19635429-19635429 on Build GRCh38
rs748965805 Chr.19: 19635412-19635412 on Build GRCh38
rs573265876 Chr.19: 19635404-19635404 on Build GRCh38
rs773920237 Chr.19: 19635418-19635418 on Build GRCh38

View Full Product Details