Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 231593828-231594093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf57
Entrez Gene ID
165100
Gene Name
chromosome 2 open reading frame 57
Gene Aliases
-
UniGene
Hs.98104
Gene Chromosome Location
Chr.2: 231592864-231594283 on Build GRCh38
Transcripts
NM_152614.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4919536 c.985C>A p.R329S Chr.2: 231593936-231593936 on Build GRCh38
3426201 c.1127T>C p.M376T Chr.2: 231594078-231594078 on Build GRCh38
4957036 c.957C>A p.D319E Chr.2: 231593908-231593908 on Build GRCh38
720263 c.897C>T p.T299T Chr.2: 231593848-231593848 on Build GRCh38
4894968 c.1093G>A p.E365K Chr.2: 231594044-231594044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770937272 Chr.2: 231593956-231593956 on Build GRCh38
rs1050527556 Chr.2: 231593853-231593853 on Build GRCh38
rs776510185 Chr.2: 231593859-231593859 on Build GRCh38
rs754657564 Chr.2: 231593924-231593924 on Build GRCh38
rs990130532 Chr.2: 231593887-231593887 on Build GRCh38
rs762488054 Chr.2: 231594069-231594069 on Build GRCh38
rs755792533 Chr.2: 231593933-231593934 on Build GRCh38
rs372291265 Chr.2: 231593990-231593990 on Build GRCh38
rs62199051 Chr.2: 231593941-231593941 on Build GRCh38
rs376770220 Chr.2: 231594001-231594001 on Build GRCh38
rs777491751 Chr.2: 231593977-231593979 on Build GRCh38
rs868367918 Chr.2: 231593986-231593986 on Build GRCh38
rs765653613 Chr.2: 231594063-231594063 on Build GRCh38
rs151146670 Chr.2: 231593923-231593923 on Build GRCh38
rs375754022 Chr.2: 231594061-231594061 on Build GRCh38
rs777214430 Chr.2: 231594057-231594057 on Build GRCh38
rs200048306 Chr.2: 231593912-231593912 on Build GRCh38
rs375509340 Chr.2: 231593936-231593936 on Build GRCh38
rs146228407 Chr.2: 231594000-231594000 on Build GRCh38
rs755461995 Chr.2: 231593996-231593996 on Build GRCh38
rs147653537 Chr.2: 231593940-231593940 on Build GRCh38
rs933082998 Chr.2: 231594020-231594020 on Build GRCh38
rs760869633 Chr.2: 231594055-231594055 on Build GRCh38
rs777111398 Chr.2: 231594006-231594006 on Build GRCh38
rs201004143 Chr.2: 231594015-231594015 on Build GRCh38
rs748329657 Chr.2: 231594035-231594035 on Build GRCh38
rs544459786 Chr.2: 231593963-231593963 on Build GRCh38
rs763127595 Chr.2: 231593868-231593868 on Build GRCh38
rs776264113 Chr.2: 231593962-231593962 on Build GRCh38
rs183366634 Chr.2: 231594027-231594027 on Build GRCh38
rs202044626 Chr.2: 231594012-231594012 on Build GRCh38
rs961547476 Chr.2: 231593994-231593994 on Build GRCh38
rs772378721 Chr.2: 231594038-231594038 on Build GRCh38
rs199772589 Chr.2: 231593970-231593970 on Build GRCh38
rs764343881 Chr.2: 231593874-231593874 on Build GRCh38
rs868821829 Chr.2: 231593862-231593862 on Build GRCh38
rs762033541 Chr.2: 231593971-231593971 on Build GRCh38
rs747769535 Chr.2: 231593893-231593895 on Build GRCh38
rs530096172 Chr.2: 231594026-231594026 on Build GRCh38
rs201807756 Chr.2: 231594014-231594014 on Build GRCh38
rs139694852 Chr.2: 231593895-231593895 on Build GRCh38
rs369891953 Chr.2: 231594040-231594040 on Build GRCh38
rs141442188 Chr.2: 231594068-231594068 on Build GRCh38
rs751621357 Chr.2: 231593888-231593888 on Build GRCh38
rs201877259 Chr.2: 231594056-231594056 on Build GRCh38
rs143308059 Chr.2: 231593947-231593947 on Build GRCh38
rs773372346 Chr.2: 231593985-231593985 on Build GRCh38
rs894252949 Chr.2: 231593942-231593942 on Build GRCh38
rs140125501 Chr.2: 231593937-231593937 on Build GRCh38
rs566726693 Chr.2: 231594029-231594029 on Build GRCh38
rs374312747 Chr.2: 231593899-231593899 on Build GRCh38
rs779081839 Chr.2: 231593991-231593991 on Build GRCh38
rs372202577 Chr.2: 231593857-231593857 on Build GRCh38
rs367910544 Chr.2: 231593856-231593856 on Build GRCh38
rs750836194 Chr.2: 231593908-231593908 on Build GRCh38
rs761055583 Chr.2: 231593988-231593988 on Build GRCh38

View Full Product Details