Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7893668-7893935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD3
Entrez Gene ID
1107
Gene Name
chromodomain helicase DNA binding protein 3
Gene Aliases
Mi-2a, Mi2-ALPHA, ZFH
UniGene
Hs.25601
Gene Chromosome Location
Chr.17: 7884764-7912757 on Build GRCh38
Transcripts
NM_001005271.2 NM_001005273.2 NM_005852.3 XM_005256427.4 XM_005256428.4 XM_005256429.4 XM_005256431.4 XM_006721423.3 XM_006721424.3 XM_006721428.3 XM_017024061.1 XM_017024062.1 XM_017024063.1 XM_017024064.1 XM_017024065.1 XM_017024066.1 XM_017024067.1 XM_017024068.1 XM_017024069.1 XM_017024070.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7266803 c.830T>C p.V277A Chr.17: 7893841-7893841 on Build GRCh38
5017884 c.822delC p.R276fs*19 Chr.17: 7893833-7893833 on Build GRCh38
1248204 c.911A>T p.K304M Chr.17: 7893922-7893922 on Build GRCh38
1640881 c.811C>T p.R271W Chr.17: 7893822-7893822 on Build GRCh38
4070329 c.827G>A p.R276Q Chr.17: 7893838-7893838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749330456 Chr.17: 7893838-7893838 on Build GRCh38
rs754814641 Chr.17: 7893832-7893832 on Build GRCh38
rs761263308 Chr.17: 7893856-7893856 on Build GRCh38
rs139173826 Chr.17: 7893853-7893853 on Build GRCh38
rs749526156 Chr.17: 7893776-7893776 on Build GRCh38
rs1057381598 Chr.17: 7893724-7893724 on Build GRCh38
rs771095708 Chr.17: 7893787-7893787 on Build GRCh38
rs775654737 Chr.17: 7893855-7893855 on Build GRCh38
rs201048464 Chr.17: 7893872-7893872 on Build GRCh38
rs909067461 Chr.17: 7893727-7893727 on Build GRCh38
rs750291146 Chr.17: 7893815-7893815 on Build GRCh38
rs540486635 Chr.17: 7893753-7893753 on Build GRCh38
rs754533336 Chr.17: 7893765-7893765 on Build GRCh38
rs111436114 Chr.17: 7893703-7893703 on Build GRCh38
rs773209157 Chr.17: 7893772-7893772 on Build GRCh38
rs761382967 Chr.17: 7893878-7893878 on Build GRCh38
rs750087267 Chr.17: 7893903-7893903 on Build GRCh38
rs774733248 Chr.17: 7893788-7893788 on Build GRCh38
rs757000641 Chr.17: 7893813-7893813 on Build GRCh38
rs370187934 Chr.17: 7893789-7893789 on Build GRCh38
rs765039093 Chr.17: 7893879-7893879 on Build GRCh38
rs369205747 Chr.17: 7893814-7893814 on Build GRCh38
rs746970749 Chr.17: 7893823-7893823 on Build GRCh38
rs372716915 Chr.17: 7893822-7893822 on Build GRCh38
rs941853208 Chr.17: 7893728-7893728 on Build GRCh38
rs201932618 Chr.17: 7893852-7893852 on Build GRCh38
rs938884332 Chr.17: 7893711-7893711 on Build GRCh38
rs149936335 Chr.17: 7893865-7893865 on Build GRCh38
rs916103618 Chr.17: 7893740-7893740 on Build GRCh38
rs1045249910 Chr.17: 7893766-7893766 on Build GRCh38
rs200504536 Chr.17: 7893764-7893764 on Build GRCh38
rs201043938 Chr.17: 7893755-7893755 on Build GRCh38
rs763706089 Chr.17: 7893795-7893795 on Build GRCh38
rs748236185 Chr.17: 7893756-7893756 on Build GRCh38
rs774537067 Chr.17: 7893851-7893851 on Build GRCh38
rs758200919 Chr.17: 7893818-7893818 on Build GRCh38
rs1019489085 Chr.17: 7893887-7893887 on Build GRCh38
rs753346769 Chr.17: 7893804-7893804 on Build GRCh38
rs776057008 Chr.17: 7893791-7893791 on Build GRCh38
rs377180914 Chr.17: 7893905-7893905 on Build GRCh38
rs771185332 Chr.17: 7893840-7893840 on Build GRCh38
rs777856475 Chr.17: 7893835-7893835 on Build GRCh38
rs761018482 Chr.17: 7893794-7893794 on Build GRCh38
rs201793105 Chr.17: 7893790-7893790 on Build GRCh38
rs945662265 Chr.17: 7893848-7893848 on Build GRCh38

View Full Product Details