Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 744758-745017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GEMIN4
Entrez Gene ID
50628
Gene Name
gem nuclear organelle associated protein 4
Gene Aliases
HC56, HCAP1, HHRF-1, p97
UniGene
Hs.499620
Gene Chromosome Location
Chr.17: 744421-752868 on Build GRCh38
Transcripts
NM_015721.2 XM_005256667.4 XM_005256670.4 XM_011523910.2 XM_011523911.2 XM_011523912.2 XM_011523913.2 XM_017024709.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4000261 c.3126C>T p.I1042I Chr.17: 744917-744917 on Build GRCh38
1661962 c.3034T>A p.Y1012N Chr.17: 745009-745009 on Build GRCh38
4686761 c.3127G>A p.G1043S Chr.17: 744916-744916 on Build GRCh38
4821824 c.3081G>C p.Q1027H Chr.17: 744962-744962 on Build GRCh38
3766150 c.3097C>T p.R1033C Chr.17: 744946-744946 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754509330 Chr.17: 744869-744869 on Build GRCh38
rs367581258 Chr.17: 744953-744953 on Build GRCh38
rs761471526 Chr.17: 744964-744964 on Build GRCh38
rs768202506 Chr.17: 744835-744850 on Build GRCh38
rs574891003 Chr.17: 744949-744949 on Build GRCh38
rs1045481 Chr.17: 744917-744917 on Build GRCh38
rs541615549 Chr.17: 744979-744979 on Build GRCh38
rs760102904 Chr.17: 744821-744823 on Build GRCh38
rs971993861 Chr.17: 744831-744831 on Build GRCh38
rs749129831 Chr.17: 744893-744893 on Build GRCh38
rs370905419 Chr.17: 744977-744977 on Build GRCh38
rs761135984 Chr.17: 744906-744906 on Build GRCh38
rs777866033 Chr.17: 744938-744938 on Build GRCh38
rs190024266 Chr.17: 744976-744976 on Build GRCh38
rs758656608 Chr.17: 744952-744952 on Build GRCh38
rs189266653 Chr.17: 744793-744793 on Build GRCh38
rs369306203 Chr.17: 744910-744910 on Build GRCh38
rs570053573 Chr.17: 744799-744799 on Build GRCh38
rs756067218 Chr.17: 744937-744937 on Build GRCh38
rs942099766 Chr.17: 744835-744835 on Build GRCh38
rs369662072 Chr.17: 744844-744844 on Build GRCh38
rs908735034 Chr.17: 744994-744994 on Build GRCh38
rs185329789 Chr.17: 744947-744947 on Build GRCh38
rs770999881 Chr.17: 744894-744894 on Build GRCh38
rs377283227 Chr.17: 744875-744875 on Build GRCh38
rs752286647 Chr.17: 744931-744931 on Build GRCh38
rs11099 Chr.17: 744813-744813 on Build GRCh38
rs868276318 Chr.17: 744967-744967 on Build GRCh38
rs567384937 Chr.17: 744903-744903 on Build GRCh38
rs751212548 Chr.17: 744919-744919 on Build GRCh38
rs767106378 Chr.17: 744930-744930 on Build GRCh38
rs531072805 Chr.17: 744800-744800 on Build GRCh38
rs773490623 Chr.17: 744987-744987 on Build GRCh38
rs187407430 Chr.17: 744786-744786 on Build GRCh38
rs180894698 Chr.17: 744916-744916 on Build GRCh38
rs201533499 Chr.17: 744896-744896 on Build GRCh38
rs772498763 Chr.17: 744840-744840 on Build GRCh38
rs766072309 Chr.17: 744867-744867 on Build GRCh38
rs200379895 Chr.17: 744975-744975 on Build GRCh38
rs1017331743 Chr.17: 744814-744814 on Build GRCh38
rs756162268 Chr.17: 744880-744880 on Build GRCh38
rs754237034 Chr.17: 744860-744860 on Build GRCh38
rs371944318 Chr.17: 744872-744872 on Build GRCh38
rs373429888 Chr.17: 744904-744904 on Build GRCh38
rs761225877 Chr.17: 744859-744859 on Build GRCh38
rs748132329 Chr.17: 744794-744794 on Build GRCh38
rs7813 Chr.17: 744946-744946 on Build GRCh38
rs750971533 Chr.17: 744868-744868 on Build GRCh38
rs759313133 Chr.17: 744923-744923 on Build GRCh38
rs777671907 Chr.17: 744887-744887 on Build GRCh38
rs752789638 Chr.17: 744880-744882 on Build GRCh38
rs375603122 Chr.17: 744816-744816 on Build GRCh38
rs746141568 Chr.17: 744900-744900 on Build GRCh38
rs762467853 Chr.17: 744861-744861 on Build GRCh38
rs776806514 Chr.17: 744912-744912 on Build GRCh38
rs753509291 Chr.17: 744945-744945 on Build GRCh38

View Full Product Details