Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46660743-46660990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DACT3
Entrez Gene ID
147906
Gene Name
dishevelled binding antagonist of beta catenin 3
Gene Aliases
DAPPER3, RRR1
UniGene
Hs.515490
Gene Chromosome Location
Chr.19: 46647564-46661138 on Build GRCh38
Transcripts
NM_145056.2 NR_040042.1

Gene Symbol
DACT3-AS1
Entrez Gene ID
100506068
Gene Name
DACT3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.434327
Gene Chromosome Location
Chr.19: 46660364-46677456 on Build GRCh38
Transcripts
NM_145056.2 NR_040042.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3743022 c.91C>T p.H31Y Chr.19: 46660974-46660974 on Build GRCh38
6999159 c.194A>G p.E65G Chr.19: 46660871-46660871 on Build GRCh38
5961604 c.129G>C p.Q43H Chr.19: 46660936-46660936 on Build GRCh38
6586643 c.210_227del18 p.R73_A78delRRAAAA Chr.19: 46660838-46660855 on Build GRCh38
4985584 c.143C>G p.A48G Chr.19: 46660922-46660922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1023397693 Chr.19: 46660781-46660781 on Build GRCh38
rs761664758 Chr.19: 46660794-46660794 on Build GRCh38
rs781689141 Chr.19: 46660909-46660909 on Build GRCh38
rs764020869 Chr.19: 46660773-46660773 on Build GRCh38
rs1034688467 Chr.19: 46660827-46660827 on Build GRCh38
rs750080611 Chr.19: 46660828-46660828 on Build GRCh38
rs748491879 Chr.19: 46660922-46660922 on Build GRCh38
rs765026749 Chr.19: 46660810-46660810 on Build GRCh38
rs961927512 Chr.19: 46660840-46660840 on Build GRCh38
rs1003214102 Chr.19: 46660820-46660820 on Build GRCh38
rs560978615 Chr.19: 46660891-46660891 on Build GRCh38
rs973458303 Chr.19: 46660894-46660894 on Build GRCh38
rs1011051259 Chr.19: 46660780-46660780 on Build GRCh38
rs996372572 Chr.19: 46660784-46660784 on Build GRCh38
rs558508905 Chr.19: 46660847-46660847 on Build GRCh38
rs541337082 Chr.19: 46660882-46660882 on Build GRCh38
rs571911443 Chr.19: 46660870-46660870 on Build GRCh38
rs538277627 Chr.19: 46660778-46660778 on Build GRCh38
rs1030491252 Chr.19: 46660792-46660792 on Build GRCh38
rs751017138 Chr.19: 46660879-46660879 on Build GRCh38
rs951897310 Chr.19: 46660933-46660933 on Build GRCh38

View Full Product Details