Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 31410901-31411162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRG1
Entrez Gene ID
4733
Gene Name
developmentally regulated GTP binding protein 1
Gene Aliases
NEDD3
UniGene
Hs.115242
Gene Chromosome Location
Chr.22: 31399553-31434186 on Build GRCh38
Transcripts
NM_004147.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3912654 c.363C>T p.I121I Chr.22: 31411032-31411032 on Build GRCh38
4544973 c.367G>A p.E123K Chr.22: 31411036-31411036 on Build GRCh38
5987297 c.349G>C p.D117H Chr.22: 31411018-31411018 on Build GRCh38
1033452 c.410C>T p.A137V Chr.22: 31411079-31411079 on Build GRCh38
6649310 c.375C>T p.A125A Chr.22: 31411044-31411044 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs565914906 Chr.22: 31410990-31410990 on Build GRCh38
rs202089239 Chr.22: 31411129-31411129 on Build GRCh38
rs775631441 Chr.22: 31410996-31410996 on Build GRCh38
rs754673913 Chr.22: 31410968-31410968 on Build GRCh38
rs766294568 Chr.22: 31411101-31411101 on Build GRCh38
rs1026579715 Chr.22: 31411039-31411039 on Build GRCh38
rs112487151 Chr.22: 31411083-31411083 on Build GRCh38
rs909219973 Chr.22: 31410947-31410947 on Build GRCh38
rs762949357 Chr.22: 31411023-31411023 on Build GRCh38
rs750232974 Chr.22: 31411065-31411065 on Build GRCh38
rs1038671794 Chr.22: 31411005-31411005 on Build GRCh38
rs762051785 Chr.22: 31411049-31411049 on Build GRCh38
rs113312163 Chr.22: 31411082-31411082 on Build GRCh38
rs149631542 Chr.22: 31410979-31410979 on Build GRCh38
rs1036652122 Chr.22: 31410983-31410983 on Build GRCh38
rs771375127 Chr.22: 31410976-31410976 on Build GRCh38
rs770257756 Chr.22: 31410991-31410991 on Build GRCh38
rs970124267 Chr.22: 31411103-31411103 on Build GRCh38
rs960578637 Chr.22: 31411133-31411133 on Build GRCh38
rs768732335 Chr.22: 31411033-31411033 on Build GRCh38
rs752237408 Chr.22: 31411111-31411111 on Build GRCh38
rs1016387037 Chr.22: 31411071-31411071 on Build GRCh38
rs200096188 Chr.22: 31411040-31411040 on Build GRCh38
rs774504553 Chr.22: 31411034-31411034 on Build GRCh38
rs572535013 Chr.22: 31410972-31410972 on Build GRCh38
rs760648021 Chr.22: 31411095-31411095 on Build GRCh38
rs901759500 Chr.22: 31411032-31411032 on Build GRCh38
rs200130318 Chr.22: 31411051-31411051 on Build GRCh38
rs778642133 Chr.22: 31410970-31410970 on Build GRCh38
rs554460015 Chr.22: 31411111-31411111 on Build GRCh38
rs754481708 Chr.22: 31411121-31411121 on Build GRCh38
rs950555434 Chr.22: 31411046-31411046 on Build GRCh38
rs997408026 Chr.22: 31411038-31411038 on Build GRCh38
rs889504257 Chr.22: 31411106-31411106 on Build GRCh38
rs144674118 Chr.22: 31411056-31411056 on Build GRCh38

View Full Product Details