Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150847974-150848201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRGM
Entrez Gene ID
345611
Gene Name
immunity related GTPase M
Gene Aliases
IFI1, IRGM1, LRG-47, LRG47
UniGene
Hs.519680
Gene Chromosome Location
Chr.5: 150846523-150902402 on Build GRCh38
Transcripts
NM_001145805.1 XM_011537641.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4960606 c.31G>A p.A11T Chr.5: 150848154-150848154 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777270209 Chr.5: 150848105-150848105 on Build GRCh38
rs749790081 Chr.5: 150848131-150848131 on Build GRCh38
rs1020331668 Chr.5: 150848155-150848155 on Build GRCh38
rs917155918 Chr.5: 150848050-150848050 on Build GRCh38
rs992774867 Chr.5: 150848047-150848047 on Build GRCh38
rs920690576 Chr.5: 150848081-150848081 on Build GRCh38
rs759880876 Chr.5: 150848052-150848052 on Build GRCh38
rs180802994 Chr.5: 150848174-150848174 on Build GRCh38
rs763474099 Chr.5: 150848130-150848130 on Build GRCh38
rs146896406 Chr.5: 150848089-150848089 on Build GRCh38
rs552962547 Chr.5: 150848026-150848026 on Build GRCh38
rs776333041 Chr.5: 150848036-150848036 on Build GRCh38
rs754653427 Chr.5: 150848060-150848060 on Build GRCh38
rs1040621750 Chr.5: 150848133-150848133 on Build GRCh38
rs892688891 Chr.5: 150848141-150848141 on Build GRCh38
rs578253387 Chr.5: 150848030-150848030 on Build GRCh38
rs755519295 Chr.5: 150848090-150848090 on Build GRCh38
rs889311877 Chr.5: 150848101-150848101 on Build GRCh38
rs1050459732 Chr.5: 150848092-150848092 on Build GRCh38
rs764528929 Chr.5: 150848150-150848150 on Build GRCh38
rs958519900 Chr.5: 150848037-150848037 on Build GRCh38
rs757815291 Chr.5: 150848148-150848148 on Build GRCh38
rs9637870 Chr.5: 150848053-150848053 on Build GRCh38

View Full Product Details